Merck
  • Risks of harms using antifibrinolytics in cardiac surgery: systematic review and network meta-analysis of randomised and observational studies.
Risks of harms using antifibrinolytics in cardiac surgery: systematic review and network meta-analysis of randomised and observational studies.
BMJ (Clinical research ed.) (2012-09-13)
Brian Hutton, Lawrence Joseph, Dean Fergusson, C David Mazer, Stan Shapiro, Alan Tinmouth
ABSTRACT
To estimate the relative risks of death, myocardial infarction, stroke, and renal failure or dysfunction between antifibrinolytics and no treatment following the suspension of aprotinin from the market in 2008 for safety reasons and its recent reintroduction in Europe and Canada. Systematic review and network meta-analysis. A Cochrane review of antifibrinolytic treatments was chosen as the starting point for this systematic review. Medline, Embase, and the Cochrane register of trials were searched with no date restrictions for observational evidence. Propensity matched or adjusted observational studies with two or more of the interventions of interest (aprotinin, tranexamic acid, epsilon-aminocaproic acid, and no treatment) that were carried out in patients undergoing cardiac surgery. Network meta-analysis was used to compare treatments, and odds ratios with 95% credible intervals were estimated. Meta-analyses were carried out for randomised controlled trials alone and for randomised controlled trials with observational studies. 106 randomised controlled trials and 11 observational studies (43,270 patients) were included. Based on the results from analysis of randomised controlled trials, tranexamic acid was associated on average with a reduced risk of death compared with aprotinin (odds ratio 0.64, 95% credible interval 0.41 to 0.99). When observational data were incorporated, comparisons showed an increased risk of mortality with aprotinin on average relative to tranexamic acid (odds ratio 0.71, 95% credible interval 0.50 to 0.98) and epsilon-aminocaproic acid (0.60, 0.43 to 0.87), and an increased risk of renal failure or dysfunction on average relative to all comparators: odds ratio 0.66 (95% credible interval 0.45 to 0.88) compared with no treatment, 0.66 (0.48 to 0.91) versus tranexamic acid, and 0.65 (0.45 to 0.88) versus epsilon-aminocaproic acid. Although meta-analyses of randomised controlled trials were largely inconclusive, inclusion of observational data suggest concerns remain about the safety of aprotinin. Tranexamic and epsilon-aminocaproic acid are effective alternatives that may be safer for patients.
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Supelco
Aminocaproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)
Sigma-Aldrich
6-Aminocaproic acid, ≥99%, BioUltra