Skip to Content
Merck
All Photos(5)

Key Documents

A7824

Sigma-Aldrich

6-Aminocaproic acid

BioUltra, ≥99%

Synonym(s):

ε-Aminocaproic acid, 6-Aminohexanoic acid, EACA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)5CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
906872
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

product line

BioUltra

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

207-209 °C (dec.) (lit.)

solubility

H2O: 0.5 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.001%
NH4+: ≤0.05%
Na: ≤0.02%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

room temp

SMILES string

NCCCCCC(O)=O

InChI

1S/C6H13NO2/c7-5-3-1-2-4-6(8)9/h1-5,7H2,(H,8,9)

InChI key

SLXKOJJOQWFEFD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

404.6 - 408.2 °F

Flash Point(C)

207 - 209 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tijana Jovanovic-Talisman et al.
Methods in cell biology, 122, 379-393 (2014-05-27)
In vitro mimics of cellular machines have been recently engineered and utilized to investigate processes within cells. These devices can provide novel insights into biological mechanisms and have the potential to improve biotechnological processes such as separation. In particular, several
Robert A Gaultney et al.
PloS one, 8(9), e75643-e75643 (2013-10-03)
The causative agent of Lyme disease, Borrelia burgdorferi, codes for several known fibronectin-binding proteins. Fibronectin a common the target of diverse bacterial pathogens, and has been shown to be essential in allowing for the development of certain disease states. Another
Weining Bian et al.
Nature protocols, 4(10), 1522-1534 (2009-10-03)
This protocol describes a cell/hydrogel molding method for precise and reproducible biomimetic fabrication of three-dimensional (3D) muscle tissue architectures in vitro. Using a high aspect ratio soft lithography technique, we fabricate polydimethylsiloxane (PDMS) molds containing arrays of mesoscopic posts with
Alastair Khodabukus et al.
Toxicological sciences : an official journal of the Society of Toxicology, 176(1), 124-136 (2020-04-16)
Traditional serum biomarkers used to assess skeletal muscle damage, such as activity of creatine kinase (CK), lack tissue specificity and sensitivity, hindering early detection of drug-induced myopathies. Recently, a novel four-factor skeletal muscle injury panel (MIP) of biomarkers consisting of
Brian Hutton et al.
BMJ (Clinical research ed.), 345, e5798-e5798 (2012-09-13)
To estimate the relative risks of death, myocardial infarction, stroke, and renal failure or dysfunction between antifibrinolytics and no treatment following the suspension of aprotinin from the market in 2008 for safety reasons and its recent reintroduction in Europe and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service