Skip to Content
Merck
All Photos(1)

Key Documents

1548134

USP

Potassium bicarbonate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Potassium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
Beilstein:
4535309
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

potassium bicarbonate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[K+].OC([O-])=O

InChI

1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

TYJJADVDDVDEDZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Potassium bicarbonate USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Potassium Bicarbonate Effervescent Tablets for Oral Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Wood
Nutrition reviews, 52(8 Pt 1), 278-280 (1994-08-01)
A recent study of postmenopausal women has shown that the administration of 60-120 mmol potassium bicarbonate can neutralize net acid excretion and significantly lower urinary calcium (Ca). The changes in urinary calcium loss were sufficient to improve calcium balance and
Federica Sangiuolo et al.
The European respiratory journal, 45(2), 483-490 (2014-12-17)
In idiopathic pulmonary fibrosis (IPF), lung accumulation of excessive extracellular iron and macrophage haemosiderin may suggest disordered iron homeostasis leading to recurring microscopic injury and fibrosing damage. The current study population comprised 89 consistent IPF patients and 107 controls. 54
Simon P Jochems et al.
Journal of virology, 89(1), 751-762 (2014-10-31)
Natural hosts of simian immunodeficiency virus (SIV), such as African green monkeys (AGMs), do not progress to AIDS when infected with SIV. This is associated with an absence of a chronic type I interferon (IFN-I) signature. It is unclear how
Fumiyuki Takasaki et al.
Dalton transactions (Cambridge, England : 2003), 44(2), 645-652 (2014-11-12)
The speciation of zirconium complexes in ammonium zirconium carbonate (AZC) solutions should be further studied in order to develop high-performance AZC complexes for use in elaborate industrial materials. Thus, the speciation and the structure of various zirconium species in ammonium
Wenchao Sheng et al.
Nature communications, 6, 5848-5848 (2015-01-09)
The hydrogen oxidation/evolution reactions are two of the most fundamental reactions in distributed renewable electrochemical energy conversion and storage systems. The identification of the reaction descriptor is therefore of critical importance for the rational catalyst design and development. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service