1493000
USP
Pamoic acid
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1,1′-Methylene-bis(2-hydroxy-3-naphthoic acid), 4,4′-Methylenebis(3-hydroxy-2-naphthoic acid), Embonic acid
About This Item
grade
pharmaceutical primary standard
API family
cycloguanil, oxantel
manufacturer/tradename
USP
mp
≥300 °C (dec.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
OC(=O)c1cc2ccccc2c(Cc3c(O)c(cc4ccccc34)C(O)=O)c1O
InChI
1S/C23H16O6/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29/h1-10,24-25H,11H2,(H,26,27)(H,28,29)
InChI key
WLJNZVDCPSBLRP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Also, for use with USP monograph such as Pyrantel Pamoate.
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service