Skip to Content
Merck
All Photos(1)

Key Documents

G006152

LC/MS (TOF) Analysis of 2C Amine Drugs of Abuse on Ascentis® Express HILIC

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express HILIC, 5 cm x 2.1 mm I.D., 2.7 μm particles (53934-U)
mobile phase: 2 mM ammonium formate in 90:10 (v:v) acetonitrile:water
flow rate: 0.4 mL/min
pressure: 943 psi (65 bar)
column temp.: 35 °C
detector: TOF/MS
injection: 2 μL
sample: 300 ng/mL in acetonitrile

suitability

application for HPLC

application(s)

forensics and toxicology

Analysis Note

The 2C amines are a family of illicit, psychedelic phenethylamines. The term "2C" refers to the two carbon atoms between the benzene ring and the amino group in the parent molecular structure. A rapid separation of four 2C amine compounds is shown here on an Ascentis Express HILIC column with TOF-MS detection. Cerilliant CRMs were used to ensure reliable MS identification and quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    (R)-(-)-Bromo-DragonFLY hydrochloride solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    2,5-Dimethoxy-4-(n)-propylthiophenethylamine hydrochloride solution, 1.0 mg/mL in acetonitrile: water (9:1) (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    4-Bromo-2,5-dimethoxyphenethylamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    2C-B-FLY hydrochloride solution, 1 mg/mL in acetonitrile: water (9:1), ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service