Skip to Content
Merck
All Photos(1)

Key Documents

59172

Supelco

SUPELCOSIL Suplex pKb-100 (5 µm) HPLC Columns

L × I.D. 25 cm × 10 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOSIL Suplex pKb-100 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm

Agency

suitable for USP L68

feature

endcapped: no

manufacturer/tradename

SUPELCOSIL

extent of labeling

12.5% carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 10 mm

surface area

170 m2/g

surface coverage

surface coverage 3.4 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

amide, alkyl phase

particle size

5 μm

pore size

120 Å

pH range

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Suplex pKb-100 columns feature the same bonded phase functionality as SUPELCOSIL LC-ABZ columns. These specially deactivated columns differ in that Suplex pKb-100 is not endcapped, while SUPELCOSIL LC-ABZ is endcapped. The absence of the end-capping reagent results in better performance from Suplex pKb-100 for the strongest basic compounds, while LC-ABZ is preferred when the sample also contains acids and/or zwitterions.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service