Key Documents
58539-U
Supelco
SSI TC-20 Tubing Cutter
AC/DC input 110 V / 220 V AC, 50-60 Hz (voltage selectable), CE compliant
Synonym(s):
Tubing Cutter Tool
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.
feature
CE compliant
General description
The SSI Model TC-20 electric stainless steel tube cutter assures a zero dead volume connection. Because the tubing is held securely in a clamp vise on the vertical swing arm, a square cut is produced when the swing arm is lowered against the abrasive cutting wheel, which produces a finished end. No lubricant or cutting fluid is required. The unit, which is CE marked, will cut most common tubing used in chromatography. Tubing with 1/16 in., 1/8 in., and 1/4 in. outside diameter, can be smooth-cut and dressed without distortion.
The precision ground dressing tool for the 1/16 in. O.D. tubing is included and is attached directly to the swing arm: it cannot be misplaced or lost. A dressing tool (deburring tool - Cat. No. 58804) for 1/8 in. diameter can be ordered separately.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
SSI is a trademark of Scientific Systems, Inc.
recommended
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Questions
  1. What is the maximum OD of SS Pipe that this can cut? We need to cut 3/8" OD .035 wall 316 SS Pipe at both flat & a 45 degree. Can this machine cut at a 45 degree angle?
    1 answer
    1. SSI TC-20 Tubing Cutter, product 58539-U, has a cutting arm that provides a square only (flat) cut of 1/16, 1/8 and 1/4 inch (1.6, 3.2 and 6.4 mm (maximum)) OD stainless steel tubing with ID down to 0.008 inches (0.2 mm).
      Helpful?