57058
Supelclean™ ENVI-Florisil® SPE Tubes
PTFE frit, bed wt. 500 mg, volume 3 mL, pk of 54
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115712
Recommended Products
material
PTFE frit
product line
Supelclean™
composition
bed wt., 500 mg
packaging
pk of 54
technique(s)
solid phase extraction (SPE): suitable
volume
3 mL
matrix
ENVI-Florisil® magnesium silicate
matrix active group
Florisil phase
diameter
100-200 mesh
application(s)
food and beverages
separation technique
normal phase
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Magnesium Silicate
Retention Mechanism: Normal phase or Adsorption
Sample Matrix Compatibility: Organic solutions
Retention Mechanism: Normal phase or Adsorption
Sample Matrix Compatibility: Organic solutions
- Mesh: 100/200; Available with PTFE or stainless steel frits
- Tested for US Environmental Protection Agency (EPA) Contract Laboratory Program (CLP) statement of work for pesticides
- Highly polar material that strongly adsorbs to polar compounds from nonpolar matrices under normal phase conditions
- Typical applications include alcohols, aldehydes, amines, herbicides, pesticides, PCBs, ketones, nitro compounds, organic acids, and phenols
Legal Information
Florisil is a registered trademark of U.S. Silica Company
Supelclean is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service