21028
GC Stationary Phase
phase Carbowax 1540, pkg of 50 g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151817
Recommended Products
packaging
pkg of 50 g
parameter
50-175 °C temp. range
technique(s)
gas chromatography (GC): suitable
matrix active group
Carbowax 1540 phase
solubility
chloroform: soluble
column type
packed GC
Looking for similar products? Visit Product Comparison Guide
General description
GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients. The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are few parameters that should be considered before choosing a stationary phase. Carbowax 1450 was also known as Carbowax 1540, it is a water-soluble solid wax with a melting temperature range of 43-46°C. It is suitable for cytological applications.
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.
Application
Carbowax 1540 stationary phase may be used for studying the correlation between retention index and Van der Waals′ volume in homologous series of n-alcohols, n-aldehydes and esters.
Kit Components Only
Product No.
Description
- Carbowax® 1540
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
281.5 °F - closed cup
Flash Point(C)
138.6 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gary Gill
Stationary Phases in Gas Chromatography (2012)
Correlations between Van der Waals' volume and retention index general equation applicable to different homologous series.
Calixto, F. Saura, and A. Garcia Raso.
Chromatographia, 14, 596-598 (1981)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service