20053
ORBO™ 615 Amberlite™ XAD®-7 (15/50), 60/30 mg
W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
47131900
Recommended Products
material
W,W,W separators
product line
Amberlite™
ORBO™
composition
Bed A, 60 mg
Bed B, 30 mg
packaging
pkg of 50 ea
manufacturer/tradename
ORBO™ 615
technique(s)
active air sampling: suitable
O.D. × L
6 mm × 75 mm
matrix
XAD-7
particle size
15-50 mesh
application(s)
air monitoring
environmental
industrial hygiene
General description
- ORBO™ 615 Amberlite™ XAD®-7 (15/50), 60/30 mg solvent desorption tube is designed for gas and vapor sampling.
- It contains two beds of the same selective adsorbent separated by glass wool or foam.
- Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
Legal Information
Amberlite is a trademark of DuPont de Nemours, Inc.
ORBO is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service