Skip to Content
Merck
All Photos(1)

Key Documents

Z763624

Breathe Easier sealing membrane for multiwell plates

pack of 100 ea

Synonym(s):

breathable sealing membrane, multiwell sealing film, plate sealers, plate sealing film, sealing film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

material

white Rayon plate (non-woven)
white wells

sterility

sterile

feature

piercable

packaging

pack of 100 ea

manufacturer/tradename

Diversified Biotech BERM-2000

size

96 wells

color

white

General description

Gas-Permeable Sealing Membrane - A Fresh Breath for Cell Growth Technology!

Breathe-Easier sealing membrane offers an exceedingly high rate of gas exchange to allow maximum cell growth rate. When adhered to a microtiter plate, the 5.5 mil thick membrane insures a uniform rate of gas exchange for all wells, while sustaining a contaminant-free environment within the well. Conventional microtiter lids disrupt uniform gas exchange from well to well by favoring those wells along the perimeter of the plate.

With Breathe-Easier, sterility is achieved using gamma irradiation. Unlike ethylene oxide sterilization, this sterilization method assures that no cytotoxins are introduced into the membranes. Breathe Easier will not inhibit fibroblast cell growth or division, and the membrane is FDA approved for blood and cell contact in wond healing. The acrylic adhesive exhibits excellent bonding characteristics with all different plastic microtiter plates.

  • High Gas Permeability- Rapid and uniform transmission of oxygen and exchange of carbon dioxide to stimulate cell growth. Water vapor transmission across membrane is 4.2 kg/m2/24 hours.
  • Non-Cytotoxic and Sterile- Membranes are gamma irradiated to assure sterility. Membranes are free of cytotoxins.
  • Thermal & Chemical Stability- Resists a majority of caustic agents and organic solvents as well as exposure to temperatures from -20 C to +80 C.
  • Simple Specimen Retrieval- Samples can be removed from plates with Breathe-Easier in place since membranes can be pierced with pipette tips.
  • Practically Designed & User-Friendly- Breathe-Easier format is 6 x 3.5 in. rectangles that fit 96-well plates, bio-blocks and tissue culture plates. Paper release liner is scored for easy application of membranes to plates.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin I Yoo et al.
Nature communications, 11(1), 5459-5459 (2020-10-31)
Biocontainment systems are needed to neutralize genetically modified organisms (GMOs) that pose ecological threats outside of controlled environments. In contrast, benign selection markers complement GMOs with reduced fitness. Benign selection agents serve as alternatives to antibiotics, which are costly and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service