Skip to Content
Merck
All Photos(1)

Key Documents

Z717177

ART® self-sealing barrier pipette tips

ART 20, volume range 0.5-20 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
0.5—20 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2149

material

colorless
colorless tip

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

pack of 960 ea (10 racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 2149

technique(s)

PCR: suitable

maximum volume

20 μL

volume range

0.5-20 μL

volume

0.5—20 μL

compatibility

for use with

  • Eppendorf Research Pro .5-10 μl
  • Eppendorf Response 4850 .5-10 μl
  • Eppendorf Reference/Ultra Micro 2-20 μl
  • Eppendorf Titerman 4908 .5-10 μl
  • Nichiryo BenchMate .5-10 μl
  • Rainin Pipet Plus 2.5-20 μl
  • Rainin Pipetman 2.5-20 μl
  • Transferpette Multi-Channel 2.20 μl

Looking for similar products? Visit Product Comparison Guide

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service