Skip to Content
Merck
All Photos(4)

Documents

WH0011137M1

Sigma-Aldrich

Monoclonal Anti-PWP1 antibody produced in mouse

clone 3A11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-IEFSSP9502, Anti-PWP1 homolog (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PWP1(11137)

Immunogen

PWP1 (AAH01652, 1 a.a. ~ 501 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MNRSRQVTCVAWVRCGVAKETPDKVELSKEEVKRLIAEAKEKLQEEGGGSDEEETGSPSEDGMQSARTQARPREPLEDGDPEDDRTLDDDELAEYDLDKYDEEGDPDAETLGESLLGLTVYGSNDQDPYVTLKDTEQYEREDFLIKPSDNLIVCGRAEQDQCNLDVHVYNQEEDSFYVHHDILLSAYPLSVEWLNFDPSPDDSTGNYIAVGNMTPVIEVWDLDIVDSLEPVFTLGSKLSKKKKKKGKKSSSAEGHTDAVLDLSWNKLIRNVLASASADNTVILWDMSLGKPAASLAVHTDKVQTLQFHPFEAQTLISGSYDKSVALYDCRSPDESHRMWRFSGQIERVTWNHFSPCHFLASTDDGFVYNLDARSDKPIFTLNAHNDEISGLDLSSQIKGCLVTASADKYVKIWDILGDRPSLVHSRDMKMGVLFCSSCCPDLPFIYALRGQKEGLRVWDISTVSSVNEAFGRRERLVLGSARNSSISGPFGSRSSDTPMES

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service