Skip to Content
Merck

V0631

Vortex adapter for microcentrifuge tubes

capacity, 8 tubes

Synonym(s):

370800008, F37080-0008, agitator adapter, stirrer adapter, stirrer adapter for tubes, vortex adapter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.73
UNSPSC Code:
41121702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


feature

capacity 8 tubes

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F37080-0008

technique(s)

cryopreservation: suitable

General description

For use with any vortex mixer equipped with a rubber pad or test tube cup. The device holds 1.5 mL tubes in a disk mounted on a shaft. A sliding plate holds down the tube caps when in use. Provides rapid and complete mixing of many samples at once.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library