SRP5177
CREB1 (1-280), GST tagged from rat
recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
Creb
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
rat
recombinant
expressed in E. coli
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~59 kDa
NCBI accession no.
application(s)
cell analysis
shipped in
dry ice
storage temp.
−70°C
Gene Information
rat ... Creb1(81646)
General description
CREB1 is a member of the cAMP responsive element binding protein family of transcription factors that are critical mediators of gene expression in response to extracellular signals and are essential regulators of adaptive behavior and long-term memory formation. Activation of CREB1 by protein kinase A-mediated phosphorylation has been implicated in the survival of mammalian cells. Mice lacking CREB in the central nervous system during development show extensive apoptosis of postmitotic neurons. CREB binds as homo- and heterodimers to promoters containing CRE and activator protein 1 (AP-1) sites to alter target-gene expression).
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Olga Valverde et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 29(6), 1122-1133 (2004-03-19)
The transcription factor cAMP-responsive element binding protein (CREB) has been shown to regulate different physiological responses including drug addiction and emotional behavior. Molecular changes including adaptive modifications of the transcription factor CREB are produced during drug dependence in many regions
Theo Mantamadiotis et al.
Nature genetics, 31(1), 47-54 (2002-04-23)
Control of cellular survival and proliferation is dependent on extracellular signals and is a prerequisite for ordered tissue development and maintenance. Activation of the cAMP responsive element binding protein (CREB) by phosphorylation has been implicated in the survival of mammalian
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service