Skip to Content
Merck
All Photos(2)

Key Documents

SML3612

Sigma-Aldrich

Ganoderic acid D

≥95% (HPLC)

Synonym(s):

(7β)-7-Hydroxy-3,11,15,23-tetraoxolanost-8-en-26-oic acid, GAD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H42O7
CAS Number:
Molecular Weight:
514.65
MDL number:
UNSPSC Code:
12352106
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O[C@H]1C[C@@H]2[C@](CCC(=O)C2(C)C)(C3=C1[C@]4([C@@]([C@H](CC4=O)[C@@H](CC(=O)CC(C)C(=O)O)C)(CC3=O)C)C)C

InChI key

YTVGSCZIHGRVAV-IYAQLQCNSA-N

Biochem/physiol Actions

Ganoderic acid D is one of major triterpenes isolated from Ganoderma lucidum that exhibit potent anticancer properties. Ganoderic acid D inhibits the energy reprogramming of colon cancer cells including glucose uptake, lactate, and pyruvate and acetyl-coenzyme production through SIRT3 upregulation via acetylated cyclophilin D inhibition.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhendong Liu et al.
European journal of pharmacology, 824, 72-77 (2018-01-29)
Ganoderic acid D (GAD) is a highly oxygenated tetracyclic triterpenoid. This study aims to assess the effects of GAD on the energy metabolism of colon cancer through the regulation of SIRT3 expression and whether this effect is related to acetylated
Ruo-Lin Zhao et al.
Journal of ethnopharmacology, 210, 287-295 (2017-09-09)
Ganoderma lucidum (GL) is an oriental medical fungus, which was used to prevent and treat many diseases. Previously, the effective compounds of Ganoderma lucidum extract (GLE) were extracted from two kinds of GL, [Ganoderma lucidum (Leyss. Ex Fr.) Karst.] and
Qing-Xi Yue et al.
Molecular & cellular proteomics : MCP, 7(5), 949-961 (2008-01-02)
Triterpenes isolated from Ganoderma lucidum could inhibit the growth of numerous cancer cell lines and were thought to be the basis of the anticancer effects of G. lucidum. Ganoderic acid D (GAD) is one of the major components in Ganoderma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service