Skip to Content
Merck
All Photos(1)

Key Documents

SML3537

Sigma-Aldrich

CK156

≥98% (HPLC)

Synonym(s):

N-tert-Butyl-7,10-dioxa-13,17,18,21-tetraazatetracyclo[12.5.2.1²,⁶.0¹⁷,²⁰]docosa-1(20),2,4,6(22),14(21),15,18-heptaene-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N5O3
Molecular Weight:
395.45
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C)(C)NC(C(C=C1)=C(OCCOCCNC2=N3)C=C1C4=C3N(C=C2)N=C4)=O

InChI

1S/C21H25N5O3/c1-21(2,3)25-20(27)15-5-4-14-12-17(15)29-11-10-28-9-7-22-18-6-8-26-19(24-18)16(14)13-23-26/h4-6,8,12-13H,7,9-11H2,1-3H3,(H,22,24)(H,25,27)

Biochem/physiol Actions

CK156 is a potent and selective DRAK1 inhibitor (IC50 = 49 nM by cell-free radiometric assay, IC50 = 181 nM by cellular NanoBRET, KD = 21 nM by ITC, selectivity by a 468-kinase panel) with good selectivity over DRAK2 (61% inhibition at 1 μM, IC50 = 8 μM by NanoBRET).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service