Skip to Content
Merck
All Photos(1)

Key Documents

SML3419

Sigma-Aldrich

Koumine

≥95% (HPLC)

Synonym(s):

(3S,4S,4aR,6R,11bS,11cS)-11c-Ethenyl-1,2,3,4,4a,5,6,11c-octahydro-2-methyl-6,4, CID 91895267

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O
CAS Number:
Molecular Weight:
306.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D 295 to 245°

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

InChI key

VTLYEMHGPMGUOT-MKWZCHDOSA-N

Biochem/physiol Actions

Koumine is a major indole alkaloid isolated from poisonous plant Gelsemium elegans that exhibits anti-inflammatory and analgesic effects in inflammatory and neuropathic pain models. It appears that koumine ameliorate chronic constriction injury induced neurotic pain through induction of autophagy and the inhibition apoptosis in the spinal cord of rodents. Koumine protects IPEC-J2 cells from hydrogen peroxide induced oxidative stress and apoptosis through decreasing reactive oxygen species (ROS) production.
Major indole alkaloid isolated from poisonous plant Gelsemium elegans that exhibits anti-inflammatory and analgesic effects

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhihang Yuan et al.
International journal of molecular sciences, 20(3) (2019-02-14)
Medicinal herbal plants have been commonly used for intervention in different diseases and improvement of health worldwide. Koumine, an alkaloid monomer found abundantly in Gelsemium plants, can be effectively used as an antioxidant. The purpose of this study was to
Gui-Lin Jin et al.
Frontiers in pharmacology, 9, 989-989 (2018-09-15)
Koumine, an indole alkaloid, is a major bioactive component of Gelsemium elegans. Previous studies have demonstrated that koumine has noticeable anti-inflammatory and analgesic effects in inflammatory and neuropathic pain (NP) models, but the mechanisms involved are not well understood. This
Qiao Ye et al.
PloS one, 14(2), e0212231-e0212231 (2019-02-13)
Koumine is a component of the Chinese medicinal herb Gelsemium elegans and is toxic to vertebrates. We used the ciliate Tetrahymena thermophila as a model to evaluate the toxic effects of this indole alkaloid in eukaryotic microorganisms. Koumine inhibited T.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service