Skip to Content
Merck
All Photos(1)

Key Documents

SML3048

Sigma-Aldrich

Tetrandrine

≥98% (HPLC)

Synonym(s):

(+)-Tetrandrine, (4aS,16aS)-3,4,4a,5,16a,17,18,19-Octahydro-12,21,22,26-tetramethoxy-4,17-dimethyl-16H-1,24:6,9-dietheno-11,15-metheno-2H-pyrido[2′,3′:17,18][1,11]dioxacycloeicosino[2,3,4-ij]isoquinoline, (S,S)-Tetrandrine, NSC 77037, Tetrandrin, (1β)-6,6′,7,12-Tetramethoxy-2,2′-dimethylberbaman

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H42N2O6
CAS Number:
Molecular Weight:
622.75
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

COc1ccc2C[C@@H]3N(C)CCc4cc(OC)c(OC)c(Oc5cc6[C@H](Cc7ccc(Oc1c2)cc7)N(C)CCc6cc5OC)c34

InChI

1S/C38H42N2O6/c1-39-15-13-25-20-32(42-4)34-22-28(25)29(39)17-23-7-10-27(11-8-23)45-33-19-24(9-12-31(33)41-3)18-30-36-26(14-16-40(30)2)21-35(43-5)37(44-6)38(36)46-34/h7-12,19-22,29-30H,13-18H2,1-6H3/t29-,30-/m0/s1

InChI key

WVTKBKWTSCPRNU-KYJUHHDHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Tetrandrine is bisbenzylisoquinoline alkaloid extracted from isolated from Stephania tetrandra S Moore that exhibits numerous pharmacological activities including anti-cancer, anti-inflammatory, negative ionotropic and chronotropic effects on myocardium, immunologic and antiallergenic effects. It has been used for treatment of tuberculosis, hyperglycemia, malaria, cancer and fever. Tetrandrine is potent calcium channel blocker. Tetrandrine targets multiple signaling pathways controlling cell cycle, apoptosis. Tetrandrine is an inhibitor of efflux pumps that reversed the clarithromycin resistance phenotype of some Mycobacterium avium complex clinical isolates.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Liu et al.
Oncotarget, 7(26), 40800-40815 (2016-10-27)
Cancer is a disease caused by the abnormal proliferation and differentiation of cells governed by tumorigenic factors. Chemotherapy is one of the major cancer treatment strategies, and it functions by targeting the physiological capabilities of cancer cells, including sustained proliferation
Kai Guo et al.
Journal of investigative surgery : the official journal of the Academy of Surgical Research, 34(7), 695-702 (2019-11-07)
We aimed at investigating the effects of Dipeptidyl peptidase-4 (DPP-4) inhibitor saxagliptin (Saxa) on mouse acute lung injury (ALI)-induced by lipopolysaccharide (LPS) and the potential mechanisms. Animals were divided into four groups: control, Saxa, LPS, and LPS + Saxa. Histopathology changes of
Bhagya N et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 97, 624-632 (2017-11-05)
Tetrandrine has been known in the treatment of tuberculosis, hyperglycemia, negative ionotropic and chronotropic effects on myocardium, malaria, cancer and fever since years together. It has been known that, tetrandrine could modulate multiple signaling molecules such as kinases of cell
Melissa Menichini et al.
The Journal of antibiotics, 73(2), 128-132 (2019-10-19)
In this study we aimed to evaluate the effect of the combination of clarithromycin and four inhibitors of efflux pumps (EPIs), including berberine (BER), carbonyl cyanide m-chlorophenylhydrazone (CCCP), piperine (PIP) and tetrandrine (TET), against 12 Mycobacterium avium complex clinical isolates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service