Skip to Content
Merck
All Photos(1)

Key Documents

SML1549

Sigma-Aldrich

Ombrabulin hydrochloride

≥98% (HPLC)

Synonym(s):

(2S)-2-Amino-3-hydroxy-N-[2-methoxy-5-[(1Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenyl]-propanamide hydrochloride, AC 7700, AVE 8062A, AVE8062, Ombrabulin HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O6 · HCl
CAS Number:
Molecular Weight:
438.90
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl.NC(CO)C(=O)Nc1c(ccc(c1)C=Cc2cc(c(c(c2)OC)OC)OC)OC

InChI

1S/C21H26N2O6.ClH/c1-26-17-8-7-13(9-16(17)23-21(25)15(22)12-24)5-6-14-10-18(27-2)20(29-4)19(11-14)28-3;/h5-11,15,24H,12,22H2,1-4H3,(H,23,25);1H

InChI key

UQNRTPFLTRZEIM-UHFFFAOYSA-N

General description

Ombrabulin is a member of the combretastatin A-4 compound class and is a tubulin-depolymerising tumour vascular-disrupting compound.

Biochem/physiol Actions

Ombrabulin is a synthetic water-soluble combretastatin analog vascular disrupting agent. Ombrabulin is a tubulin polymerization inhibitor. Ombrabulin binds to the colchicine binding site of endothelial cell tubulin, inhibiting tubulin polymerization and inducing mitotic arrest and apoptosis in endothelial cells.
Ombrabulin leads to the constriction of the arteries, inhibits cell proliferation and causes detachment of endothelial cells, thereby leading to cytotoxicity. In pre-clinical studies, this compound, along with cisplatin, has shown anti-tumor activity. In tumors, it is responsible for the destruction of the vasculature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ombrabulin plus cisplatin versus placebo plus cisplatin in patients with advanced soft-tissue sarcomas after failure of anthracycline and ifosfamide chemotherapy: a randomised, double-blind, placebo-controlled, phase 3 trial.
Blay JY et al
Lancet Oncology, 16(5), 531-540 (2015)
Phase I safety, pharmacokinetic and pharmacodynamic evaluation of the vascular disrupting agent ombrabulin (AVE8062) in patients with advanced solid tumors.
Sessa C et al
Clinical Cancer Research, 19(17), 4832-4842 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service