Skip to Content
Merck
All Photos(1)

Key Documents

SML1325

Sigma-Aldrich

HS-72

≥98% (HPLC)

Synonym(s):

(S)-N-(1-propyl-1H-benzimidazol-2-yl)-1-(2-pyrazinyl)-3-Piperidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N6O
Molecular Weight:
364.44
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

HS-72 is a cell permeable, highly selective allosteric inhibitor of inducible Hsp70 (Hsp70i) over constitutively active Hsc70. HS-72 inhibits cancer cell proliferation and promotes a survival in a HER2+ mouse model of breast cancer.
HS-72 is also termed as ((S)-N-(1-propyl-1H-benzo[d]imidazol-2-yl)-1-(pyrazin-2-yl)piperidine-3-carboxamide). It is considered as a powerful anti-dengue virus (DENV) therapeutic agent. It displays anti-DENV activity in monocyte and liver cell lines. HS-72 stimulates the accumulation of cellular proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew K Howe et al.
Chemistry & biology, 21(12), 1648-1659 (2014-12-17)
Inducible Hsp70 (Hsp70i) is overexpressed in a wide spectrum of human tumors, and its expression correlates with metastasis, poor outcomes, and resistance to chemotherapy in patients. Identification of small-molecule inhibitors selective for Hsp70i could provide new therapeutic tools for cancer
An inducible heat shock protein 70 small molecule inhibitor demonstrates anti-dengue virus activity, validating Hsp70 as a host antiviral target
Howe M K, et al.
Antiviral Research, 130, 81-92 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service