Skip to Content
Merck
All Photos(1)

Key Documents

SML0848

Sigma-Aldrich

LM22A-4

≥98% (HPLC)

Synonym(s):

N1,N3,N5-Tris(2-hydroxyethyl)-1,3,5-benzenetricarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N3O6
CAS Number:
Molecular Weight:
339.34
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

N(CCO)C(=O)c1cc(cc(c1)C(=O)NCCO)C(=O)NCCO

InChI

1S/C15H21N3O6/c19-4-1-16-13(22)10-7-11(14(23)17-2-5-20)9-12(8-10)15(24)18-3-6-21/h7-9,19-21H,1-6H2,(H,16,22)(H,17,23)(H,18,24)

InChI key

RGWJKANXFYJKHN-UHFFFAOYSA-N

Application

LM22A-4 has been used as a component of dissociation medium for mouse embryonic stem cells (mESC) and human embryonic stem (hESC) aggregates.

Biochem/physiol Actions

LM22A-4 is a non-peptide and shares similarity with brain-derived neurotrophic factor (BDNF), especially in the loop2 region. It improves oligodendroglia density and favors myelin repair. LM22A-4 elevates the expression of protein kinase B (PKB or Akt) and extracellular signal-regulated kinases (ERKs). LM22A-4 elicits neuroprotection in spinal cord injury (SCI) by deregulating the cleaved-caspase-3 and favoring B-cell lymphoma 2(Bcl-2) expression.
LM22A-4 is a partial agonist of TrkB, a brain-derived neurotrophic factor (BDNF) receptor. In binding assays, LM22A-4 displaces BDNF with an IC50 of 47 nM. The compound restores TrkB phosphorylation and respiratory function in a mouse model of Rett syndrome.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TrkB Agonist LM22A-4 Increases Oligodendroglial Populations During Myelin Repair in the Brain
Nguyen HTH, et al.
Frontiers in Molecular Neuroscience, 642918-642918 (2019)
Protective effects of LM22A-4 on injured spinal cord nerves
Yu G and Wang W
International Journal of Clinical and Experimental Pathology, 8(6), 6526-6526 (2015)
Tanycyte-like cells derived from mouse embryonic stem culture show hypothalamic neural stem/progenitor cell functions
Kano M, et al.
Endocrinology, 160(7), 1701-1718 (2019)
Vasopressin-secreting neurons derived from human embryonic stem cells through specific induction of dorsal hypothalamic progenitors
Ogawa K, et al.
Scientific Reports, 8(1), 3615-3615 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service