Skip to Content
Merck
All Photos(1)

Key Documents

SMB00321

Sigma-Aldrich

Narirutin

≥98% (HPLC)

Synonym(s):

Narirutin, (2S)-7-((6-O-(6-Deoxy-a-L-mannopyranosyl)-β-D-glucopyranosyl)oxy)-2,3-dihydro-5-hydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Isonaringenin, Isonaringin, Naringenin rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32O14
CAS Number:
Molecular Weight:
580.53
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

mp

180 °C

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

OC1=CC=C([C@@H]2CC(C(C(O)=CC(O[C@@H]3O[C@H](CO[C@H]4[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O4)[C@@H](O)[C@H](O)[C@H]3O)=C5)=C5O2)=O)C=C1

InChI

1S/C27H32O14/c1-10-20(31)22(33)24(35)26(38-10)37-9-18-21(32)23(34)25(36)27(41-18)39-13-6-14(29)19-15(30)8-16(40-17(19)7-13)11-2-4-12(28)5-3-11/h2-7,10,16,18,20-29,31-36H,8-9H2,1H3/t10-,16-,18+,20-,21+,22+,23-,24+,25+,26+,27+/m0/s1

InChI key

HXTFHSYLYXVTHC-AJHDJQPGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Narirutin is a flavanone that constitutes the major flavonoids found in citrus fruits like oranges, grapefruits, and tangerines. It is a multi-potent phytochemical.

Application

Narirutin has been used as a flavonoid supplement to study its effect on the prevention of depression-like behavior and modulatory effect on the inflammatory profile in rat female offspring.

Biochem/physiol Actions

Narirutin is known to possess anti-allergic, anti-inflammatory, antioxidant, and anti-proliferative properties. It inhibits allergic eosinophilic airway inflammation in the mouse model. Narirutin also inhibits lipopolysaccharide (LPS)-induced inflammatory response by inhibiting nuclear factor-κB (NF-κB) and mitogen-activated protein kinases (MAPKs). It displays antioxidant and protective effects on attenuating alcohol-induced liver damage while co-consumed with alcohol in mice model, and can be useful in Alzheimer′s disease (AD) therapeutics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sang Keun Ha et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(10), 3498-3504 (2012-07-21)
In this study, we examined the regulatory activity of narirutin fraction from citrus peels on the production of inflammatory mediators managing acute or chronic inflammatory diseases in macrophages. Narirutin fraction inhibited the release, by lipopolysaccharide (LPS)-stimulated macrophages, of nitric oxide
Bing Zhang et al.
Molecules (Basel, Switzerland), 24(11) (2019-05-31)
A UHPLC-QQQ-MS/MS method was developed to quantify the significant constituents in Wen-Dan Decoction (WDD), a traditional Chinese medicine. Analysis of 19 compounds was conducted on an ACQUITY UPLC® BEH C18 Column (2.1 × 50 mm, 1.7 μm) using elution with
Sandipan Chakraborty et al.
International journal of biological macromolecules, 103, 733-743 (2017-05-23)
Alzheimer's disease (AD) is a complex disorder and the disease mechanism is yet to be properly characterized. Over the years, "amyloid cascade" emerges as principal pathogenic event in AD. β-Secretase 1 (BACE1) controls the rate-limiting step in amyloid beta (Aβ)
Ho-Young Park et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 55, 637-644 (2013-02-19)
This study aimed to demonstrate protective activities of the narirutin fraction from peels of Citrus unshiu against ethanol-induced hepatic damage through an animal study. Citrus narirutin fraction (CNF), contained 75% of narirutin, was obtained by an ultra-sonicated extraction and further
Lara Alexander et al.
Food chemistry, 276, 626-635 (2018-11-10)
The bitter taste of Cyclopia genistoides infusions is unacceptable to consumers, who are used to the slightly sweet taste of the herbal teas produced from other Cyclopia species. Bitter taste intensities of crude phenolic fractions of a bitter hot water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service