Skip to Content
Merck
All Photos(1)

Key Documents

SCP0145

Sigma-Aldrich

Fibrinopeptide B human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H93N19O25
Molecular Weight:
1552.56
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Glp-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg

Application

Pyroglutamated Fibrinopeptide B (Glp-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg) is related to Fibrinopeptide B (Gln-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg) derived from fibrinogen (desAA-fibrin) by thrombin activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yutaka Masuda et al.
Peptides, 23(3), 409-411 (2002-02-12)
We detected a peptide having a behavioral activity on mouse forced swimming from sera of healthy volunteers without affective and psychotic diseases. The amino acid sequence was GVNDNEEGF, which was found in the sequence of human beta-fibrinogen. The synthesized peptide
W Ruf et al.
Biochimica et biophysica acta, 965(2-3), 169-175 (1988-05-12)
Thrombin preferentially cleaves fibrinopeptides A (FPA) from fibrinogen resulting in the formation of desAA-fibrin from which most of the fibrinopeptides B (FPB) are then released with an enhanced rate. Kinetics of fibrinopeptide release from normal and dysfunctional fibrinogens were investigated
Tomas Riedel et al.
Blood, 117(5), 1700-1706 (2010-11-26)
Fibrinogen adsorption on a surface results in the modification of its functional characteristics. Our previous studies revealed that fibrinogen adsorbs onto surfaces essentially in 2 different orientations depending on its concentration in the solution: "side-on" at low concentrations and "end-on"

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service