Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502518

Sigma-Aldrich

Anti-KCNT1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KCa4.1, Potassium channel subfamily T member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 138 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNT1(57582)

General description

Anti-KCNT1 antibody detects endogenous levels of total KCNT1 protein.
The KCNT1 (potassium sodium-activated channel subfamily T member 1) gene is mapped to human chromosome 9q34.3. It is widely expressed in the brain, heart and dorsal root ganglia and well distributed in the nervous system.

Immunogen

The antiserum was produced against synthesized peptide derived from human KCNT1.

Immunogen Range: 1019-1068

Biochem/physiol Actions

Mutations in KCNT1 (potassium sodium-activated channel subfamily T member 1) is observed in different epileptic disorders including epileptic encephalopathy, nocturnal frontal lobe epilepsy and malignant migrating partial seizures in infancy. KCNT1 is associated with neural oscillation and general learning disability. It also controls pain sensation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epilepsy-Related Slack Channel Mutants Lead to Channel Over-Activity by Two Different Mechanisms.
Tang QY
Cell Reports, 9314(1), 129-139 (2016)
Characterization of two de novoKCNT1 mutations in children with malignant migrating partial seizures in infancy.
Rizzo F
Molecular and Cellular Neurosciences, 72, 54-63 (2016)
Stimulation of Slack K(+) Channels Alters Mass at the Plasma Membrane by Triggering Dissociation of a Phosphatase-Regulatory Complex.
Fleming MR
Cell Reports, 16(9), 2281-2288 (2016)
Mutations in KCNT1 cause a spectrum of focal epilepsies.
M?ller RS
Epilepsia, 56(9) (2015)
Missense mutations in the sodium-gated potassium channel gene KCNT1 cause severe autosomal dominant nocturnal frontal lobe epilepsy.
Heron SE
Nature Genetics, 44(11), 1188-1190 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service