Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200811

Sigma-Aldrich

Anti- Neurofilament 200-FITC antibody, Mouse monoclonal

clone NE14, purified from hybridoma cell culture

Synonym(s):

H-subunit, NF-H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

FITC conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

NE14, monoclonal

form

buffered aqueous solution

mol wt

200 kDa

species reactivity

pig, feline, chicken, bovine, human, mouse, guinea pig, rat

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 1:200-1:400 (4-8 ug/mL) using enzyme treated formalin-fixed, paraffin-embedded rat Cerebellum sections

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEFH(4744)

General description

Neurofilaments are type of intermediate filaments (IFs) that serve as major elements of the cytoskeleton supporting the axon cytoplasm of neuronal cells. IFs are components of most eukaryotic cells and significantly differ from other cytoskeletal elements of the cell, namely microtubules and microfilaments.

Specificity

Monoclonal Anti-Neurofilament 200, also known as Neurofilament-H or Heavy subunit, specifically recognizes the phosphorylated H tail of Neurofilament 200 and shows no reactivity on enzymatically dephosphorylated neurofilaments.2 The antibody shows reactivity with neurofilaments in the central and peripheral nervous systems from human1, pig1, mouse3, rat4, chicken3, guinea pig3, feline3 and bovine3 origin.

Immunogen

Neurofilaments purified from pig spinal cord

Application

The antibody may be used in various immunochemical techniques including Immunohistochemistry and Immunoblotting (~200 kDa).1-7

Biochem/physiol Actions

Neurofilaments undergo post-translational modifications including different levels of phosphorylation, which has been suggested to modulate their function by influencing the interaction between neurofilament and cytoplasmic organelles. Neurofilaments are built from three intertwined protofibrils of apparent molecular weights [68 (L), 160 (M) and 200 (H) kDa] which are themselves composed of two tetrameric protofilament complexes of monomeric proteins. Neurofilament 200 also known as Neurofilament heavy polypeptide (H-subunit), NF-H, NEFH or 200 kDa neurofilament protein, has an important function in mature axons that is not subserved by the two smaller neurofilament proteins.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours. Protect from prolonged exposure to light.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai Wang et al.
International wound journal, 19(8), 2000-2011 (2022-03-23)
Fasudil is reported to be effective at protecting against ischaemic diseases, and at augmenting axon growth. In this study, we aim to evaluate its efficacy in promoting flap survival and reinnervation. Ninety-two Institute of Cancer Research (ICR) mice were used
Hai Wang et al.
Journal of orthopaedic translation, 47, 74-86 (2024-07-15)
The functional recovery after peripheral nerve injury remains unsatisfactory. This study aims to perform a comprehensive evaluation of the efficacy of Fasudil Hydrochloride at treating the sciatic nerve transection injury in rats and the mechanism involved. In animal experiments, 75
Yuan Zhang et al.
Theranostics, 11(19), 9342-9357 (2021-10-15)
Background: Neuromedin B (Nmb) is implicated in the regulation of nociception of sensory neurons. However, the underlying cellular and molecular mechanisms remain unknown. Methods: Using patch clamp recording, western blot analysis, immunofluorescent labelling, enzyme-linked immunosorbent assays, adenovirus-mediated shRNA knockdown and
Yun Xie et al.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society, 28(6), 823-833 (2020-09-10)
The process of reinnervation, an important component of functional restoration after flap transfer, is understudied, making it necessary to carry out a further investigation for delineation of the exact timeline. Seventy-six Sprague-Dawley rats were used as the experimental animals. An

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service