Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200370

Sigma-Aldrich

Anti-PVRL4 antibody, Mouse monoclonal

clone N4.101.10, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Nectin-4/PVRL4 antibody produced in mouse, Monoclonal Anti-PVRL4 antibody produced in mouse, Anti-EDSS1, Anti-LNIR, Anti-Nectin-4, Anti-PRR4, Anti-PVRL4, Anti-Poliovirus receptor-related 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

N4.101.10, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

flow cytometry: 5-10 μg/test using using 1x106 MDA-MB-231 cells
immunocytochemistry: 2.5-5 μg/mL using using MDA-MB-231 cells

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PVRL4(81607)

General description

Monoclonal Anti-Nectin-4/PVRL4 (mouse IgG1 isotype) is derived from the hybridoma R2.525.2 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with human nectin4-Fc recombinant protein. Nectin-4 belongs to the family of Nectins. It forms homo-cis-dimers and then homo- or hetero-trans-dimers through the extracellular region in a Ca2+-independent manner. Nectin-4 is expressed in placenta, ductal breast carcinoma and lung adenocarcinomas. It is one of the plasma membrane components of adherens junctions.

Immunogen

human Nectin4/Fc recombinant protein

Application

Monoclonal Anti-PVRL4 antibody produced in mouse has been used in immunocytochemistry and flow cytometry.

Biochem/physiol Actions

Nectin-4 is involved in cell adhesion through trans homophilic and heterophilic interactions. Mutations in this gene are associated with ectodermal dysplasia-syndactyly syndrome type 1.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie Fabre et al.
The Journal of biological chemistry, 277(30), 27006-27013 (2002-05-16)
Nectins form a family of integral molecules that belong to the immunoglobulin superfamily. Their ectodomain is made of three Ig-like domains (V, C, C). This family comprises at least five members, namely nectin1, -2, -3, -4, and poliovirus receptor (PVR)
Nectin-4 is a new histological and serological tumor associated marker for breast cancer
Fabre-Lafay S, et al.
BMC Cancer, 7(1), 73-73 (2007)
Francesco Brancati et al.
American journal of human genetics, 87(2), 265-273 (2010-08-10)
Ectodermal dysplasias form a large disease family with more than 200 members. The combination of hair and tooth abnormalities, alopecia, and cutaneous syndactyly is characteristic of ectodermal dysplasia-syndactyly syndrome (EDSS). We used a homozygosity mapping approach to map the EDSS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service