Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401214

Sigma-Aldrich

Anti-IL12RB1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CD212, IL-12R-BETA1, IL12RB, MGC34454

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL12RB1(3594)

General description

IL12RB1 (interleukin 12 receptor subunit β1) gene is mapped to human chromosome 19p13.11. The encoded protein is a member of type I transmembrane receptors.
Rabbit polyclonal antibody raised against a full-length human IL12RB1 protein.

Immunogen

IL12RB1 (AAH29121.1, 1 a.a. ~ 381 a.a) full-length human protein.

Sequence
MEPLVTWVVPLLFLFLLSRQGAACRTSECCFQDPPYPDADSGSASGPRDLRCYRISSDRYECSWQYEGPTAGVSHFLRCCLSSGRCCYFAAGSATRLQFSDQAGVSVLYTVTLWVESWARNQTEKSPEVTLQLYNSVKYEPPLGDIKVSKLAGQLRMEWETPDNQVGAEVQFRHRTPSSPWKLGDCGPQDDDTESCLCPLEMNVAQEFQLRRRRLGSQGSSWSKWSSPVCVPPENPPQPQVRFSVEQLGQDGRRRLTLKEQPTQLELPEGCQGLAPGTEVTYRLQLHMLSCPCKAKATRTLHLGKMPYLSGAAYNVAVISSNQFGPGLNQTWHIPADTHTDGMISAHCNLRLPDSRDSPASASRVAGITGICHHTRLILYF

Biochem/physiol Actions

IL12RB1 (interleukin 12 receptor subunit β1) regulates the physiological responses to a number of diseases. IL12RB1 binds to both IL 12 (interleukin 12) and IL 23 (interleukin 23) at a common binding point 12p40-domain.Variation in the gene affects the receptor sensitivity towards the cytokines IL 12 and IL 23. Thus, the gene is associated with the diseases that are regulated by IL 12 and IL 23. Susceptibility to diseases such as tuberculosis, nontuberculous mycobacterial infection, malaria, cancer, pediatric asthma, and atopic dermatitis is related to IL12RB1.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The introduction of RNA-DNA differences underlies interindividual variation in the human IL12RB1 mRNA repertoire.
Turner AJ
Proceedings of the National Academy of Sciences of the USA, 112(50), 15414-15419 (2015)
Diverse genome-wide association studies associate the IL12/IL23 pathway with Crohn Disease.
Wang K
American Journal of Human Genetics, 84(3), 399-405 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service