Skip to Content
Merck
All Photos(1)

Key Documents

S5816

Sigma-Aldrich

Stromal Cell-Derived Factor 1α/pre-B Cell Growth Stimulating Factor from mouse

>97% (SDS-PAGE), recombinant, expressed in E. coli, powder, suitable for cell culture

Synonym(s):

SDF-1α/PBSF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Stromal Cell-Derived Factor 1α/pre-B Cell Growth Stimulating Factor from mouse, SDF-1α, recombinant, expressed in E. coli, powder, suitable for cell culture

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

>97% (SDS-PAGE)

form

powder

potency

0.15-0.6 ng/mL ED50

quality

endotoxin tested

mol wt

predicted mol wt ~8 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/μgtested (LAL test)

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Cxcl12(20315)

General description

Stromal derived factor-1 (SDF-1) is localized to adipocytes.

Application

Stromal Cell-Derived Factor 1α/pre-B Cell Growth Stimulating Factor from mouse has been used in invasion assay. It has also been used to study the SDF-1 release from chitosan-based nanoparticles.

Biochem/physiol Actions

SDF-1α/PBSF, a member of the α (CXC) chemokine family, is a chemoattractant for T lymphocytes and monocytes, but not neutrophils. SDF-1α/PBSF is highly conserved between species, with only one amino acid substitution between the human and mouse proteins. SDF-1α and SDF-1β (together also known as SDF-1α/PBSF) are encoded by a single gene and arise by alternative splicing. The two proteins are identical except for the four amino acid residues that are present in the C-terminal of SDF-1β and absent from SDF-1α..
Stromal derived factor-1 (SDF-1) is an insulin-desensitizing factor, upregulated during both starvation and obesity. Exogenic SDF-1 reduces insulin-mediated signaling and glucose uptake. SDF-1 mediates migration of cortical interneurons and endothelial progenitor cells of the bone marrow through chemotaxis.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 0.5 mg bovine serum albumin.

Preparation Note

Recombinant Mouse Stromal Cell-Derived Factor 1α (SDF-1α)/Pre-B Cell Growth Stimulating Factor (PBSF) is produced from a DNA sequence encoding the mature mouse SDF-1α protein sequence (amino acid residues 22-89) expressed in Escherichia coli. Recombinant mouse SDF-1α (68 amino acids) has a predicted molecular mass of approximately 8 kDa.

Analysis Note

The biological activity is measured by its ability to chemoattract cultured human lymphocytes and mouse BaF3 cells transfected with human CXCR4.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CXCR4 receptor positive spheroid forming cells are responsible for tumor invasion in vitro
Krohn A, et al.
Cancer Letters, 280, 65-71 (2009)
Mobilization of mesenchymal stem cells by stromal cell-derived factor-1 released from chitosan/tripolyphosphate/fucoidan nanoparticles
Huang YC, et al.
Acta Biomaterialia, 8, 1048-1056 (2012)
SDF-1 Is an Autocrine Insulin-Desensitizing Factor in Adipocytes.
Shin J, et al.
Diabetes, 67(6), 1068-1078 (2018)
SDF-1α-induced dual pairs of E-selectin/ligand mediate endothelial progenitor cell homing to critical ischemia
Liu ZJ, et al.
Scientific Reports, 6: 34416 (2016)
Yoshiki Omatsu et al.
Immunity, 33(3), 387-399 (2010-09-21)
Hematopoietic stem cells (HSCs) and their lympho-hematopoietic progeny are supported by microenvironmental niches within bone marrow; however, the identity, nature, and function of these niches remain unclear. Short-term ablation of CXC chemokine ligand (CXCL)12-abundant reticular (CAR) cells in vivo did not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service