Skip to Content
Merck
All Photos(1)

Key Documents

RAB0699

Sigma-Aldrich

Human IL34 / Interleukin-34 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 7 pg/mL
standard curve range: 7.81-250 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL34(146433)

General description

The antibody pair provided in this kit recognizes human IL-34.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin Klaeske et al.
Transplant immunology, 59, 101259-101259 (2019-11-11)
Tolerance induction following organ transplantation can be achieved by adoptive cell transfer of regulatory T-cells (Tregs) or dendritic cells (DCs). However, the target immunological profile is unknown. The present study aimed to identify an immunological profile connected to tolerance induction
Nabeel Kajihara et al.
Breast cancer (Tokyo, Japan), 27(6), 1198-1204 (2020-06-25)
Triple-negative breast cancer (TNBC) is a subtype characterized by the absence of therapeutic targets. It shows rapid progression, higher relapse, and poor prognosis, so the establishment of an effective therapeutic target is required. We focused on interleukin-34 (IL-34) that is
Thibaut Blondy et al.
Journal for immunotherapy of cancer, 8(1) (2020-06-26)
Malignant pleural mesothelioma (MPM) is a rare and aggressive cancer related to asbestos exposure. The tumor microenvironment content, particularly the presence of macrophages, was described as crucial for the development of the disease. This work aimed at studying the involvement
Hiraku Endo et al.
International immunology, 32(3), 175-186 (2019-12-24)
Ovarian cancer is the second-most lethal gynecological malignancy and the seventh-commonest cause of cancer-related death in women around the world. Most of the ovarian cancer patients are diagnosed at advanced stages and suffer from recurrence after primary cytoreductive surgery and
Veronika Kana et al.
The Journal of experimental medicine, 216(10), 2265-2281 (2019-07-28)
Microglia, the brain resident macrophages, critically shape forebrain neuronal circuits. However, their precise function in the cerebellum is unknown. Here we show that human and mouse cerebellar microglia express a unique molecular program distinct from forebrain microglia. Cerebellar microglial identity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service