Skip to Content
Merck
All Photos(2)

Key Documents

RAB0435

Sigma-Aldrich

Human SOST ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type urine
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 40 pg/mL
standard curve range: 40.96-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SOST(50964)

General description

The Human SOST (also known as sclerostin) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human SOST in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human SOST

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edyta Golembiewska et al.
BMC nephrology, 21(1), 43-43 (2020-02-09)
Vascular calcification (VC) is an independent predictor of cardiovascular disease (CVD) present in 30-70% of patients with chronic kidney disease (CKD). Copeptin is a sensitive surrogate marker of arginine vasopressin (AVP), which is involved in many pathophysiologic processes in CKD.
Annelies De Maré et al.
Journal of clinical medicine, 8(12) (2019-11-24)
Mounting evidence indicates that sclerostin, a well-known inhibitor of bone formation, may qualify as a clinically relevant biomarker of chronic kidney disease-related mineral and bone disorder (CKD-MBD), including abnormal mineral and bone metabolism and extraskeletal calcification. For this purpose, in
C Tsentidis et al.
Osteoporosis international : a journal established as result of cooperation between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA, 28(3), 945-953 (2016-10-22)
Higher levels of Dickkopf-1, which is an inhibitor of Wnt/β-catenin bone metabolic pathway, could be indicative of downregulated Wnt system, with possible lower osteoblast activation and higher osteoclast signaling in type 1 diabetes mellitus children and adolescents. Dickkopf-1 could significantly
Kuo-Chin Hung et al.
International journal of molecular sciences, 21(22) (2020-11-22)
We have previously demonstrated calcimimetics optimize the balance between osteoclastic bone resorption and osteoblastic mineralization through upregulating Wingless and int-1 (Wnt) signaling pathways in the mouse and cell model. Nonetheless, definitive human data are unavailable concerning therapeutic effects of Cinacalcet
Pamela S Hinton et al.
Bone, 96, 85-90 (2016-10-21)
We previously reported that 12months of resistance training (RT, 2×/wk, N=19) or jump training (JUMP, 3×/wk, N=19) increased whole body and lumbar spine BMD and increased serum bone formation markers relative to resorption in physically active (≥4h/wk) men (mean age:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service