Skip to Content
Merck
All Photos(1)

Key Documents

RAB0279

Sigma-Aldrich

Human IL-1 sRI ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8.19-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL1R1(3554)

General description

The Human IL-1sRI (interleukin 1 soluble receptor I) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme- linked immunosorbent assay for the quantitative measurement of human IL- 1sRI in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL-1 R1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faith Nutter et al.
Endocrine-related cancer, 21(2), 327-341 (2014-01-15)
Advanced breast cancer is associated with the development of incurable bone metastasis. The two key processes involved, tumour cell homing to and subsequent colonisation of bone, remain to be clearly defined. Genetic studies have indicated that different genes facilitate homing
Tao Gu et al.
African health sciences, 14(2), 356-363 (2014-10-17)
An early differentiation of malignant from benign solitary pulmonary nodules (SPNs) is essential for management and prognosis of lung cancer. Here we investigated whether measurement of circulating sex hormones could be useful for an early detection of malignancy among patients
Nathella Pavan Kumar et al.
The Journal of infectious diseases, 210(10), 1670-1678 (2014-06-08)
Diabetes mellitus type 2 (DM) is known to be a major risk factor for the development of active tuberculosis, although its influence on latent Mycobacterium tuberculosis infection (hereafter, "latent infection") remains poorly characterized. We examined circulating plasma cytokine levels in
Hui Wang et al.
Mediators of inflammation, 2014, 858763-858763 (2014-09-13)
Diabetic retinopathy (DR) is a major microvascular complication in diabetics, and its mechanism is not fully understood. Toll-like receptor 4 (TLR4) plays a pivotal role in the maintenance of the inflammatory state during DR, and the deletion of TLR4 eventually
Fei Zheng et al.
Mediators of inflammation, 2014, 507208-507208 (2014-07-08)
The role of the NLRP3 inflammasome in atherosclerosis remains controversial. The aim of this study was to determine whether inhibition of NLRP3 signaling by lentivirus-mediated RNA interference could reduce atherosclerosis and stabilizes plaques. We also tried to explore the mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service