Skip to Content
Merck
All Photos(1)

Key Documents

RAB0023

Sigma-Aldrich

Zinc-binding α-2 glycoprotein EIA Kit

for serum, plasma, culture supernatant and cell lysates

Synonym(s):

ZAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

species reactivity

human, rat, mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type culture supernatant(s)
sample type plasma
sample type serum
sample type cell lysate

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 21 pg/mL
standard curve range: 0.1-1000 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AZGP1(563)

General description

The ZAG Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting ZAG peptide based on the principle of Competitive Enzyme Immunoassay. The EIA-ZAG-1 detects ZAG 278aa.

Immunogen

Zinc-binding α-2 glycoprotein (ZAG), synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service