Skip to Content
Merck
All Photos(1)

Key Documents

R9400

Sigma-Aldrich

Monoclonal Anti-RANTES antibody produced in rat

clone 53405.111, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-D17S136E, Anti-RANTES, Anti-SCYA5, Anti-SIS-delta, Anti-SISd, Anti-TCP228, Anti-eoCP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

53405.111, monoclonal

form

lyophilized powder

purified by

using protein G

species reactivity

mouse

technique(s)

immunocytochemistry: 8-25 μg/mL using mouse splenocytes
neutralization: suitable

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ccl5(20304)

Related Categories

Immunogen

purified, E. coli-derived, recombinant mouse RANTES.

Biochem/physiol Actions

RANTES (Regulated upon Activation, Normal T cell Expressed and Secreted; CCL5) is a chemokine that brings T cells, monocytes, eosinophils, NK cells, dendritic cells and mast cells to the site of infection or inflammation. RANTES has an unusual kinetic profile and is secreted 3-5 days after the activation of the T cells. It also exhibits antimicrobial activity by inducing the production of nitric oxide in macrophages. The expression of RANTES is regulated mainly by JAK/STAT and NF-κB pathways.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose pH 7.4.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Appay et al.
Trends in immunology, 22(2), 83-87 (2001-04-05)
The activity of the chemokine RANTES is not restricted merely to chemotaxis. It is a powerful leukocyte activator, a feature potentially relevant in a range of inflammatory disorders. RANTES has attracted attention because it can potently suppress and, in some
Alan M Krensky et al.
Nature clinical practice. Nephrology, 3(3), 164-170 (2007-02-27)
Chemokines (chemoattractant cytokines) are fundamental regulators of immune cell movement from the bloodstream into tissues. Regulating expression of chemokines might, therefore, alleviate inflammation in autoimmune diseases and transplant rejection, or augment immune responses in cancer and immunodeficiency. RANTES (regulated upon
F Villalta et al.
Infection and immunity, 66(10), 4690-4695 (1998-09-24)
This paper describes a new role for the cysteine-cysteine (CC) chemokines RANTES, MIP-1alpha, and MIP-1beta on human macrophage function, which is the induction of nitric oxide (NO)-mediated trypanocidal activity. In a previous report, we showed that RANTES, MIP-1alpha and MIP-1beta
The unexpected pleiotropic activities of RANTES.
Jay A Levy
Journal of immunology (Baltimore, Md. : 1950), 182(7), 3945-3946 (2009-03-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service