Skip to Content
Merck
All Photos(1)

Key Documents

R7638

Sigma-Aldrich

RPMI-1640 Medium

Dutch Modification, with sodium bicarbonate and HEPES, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

pH

>7.2

components

sodium pyruvate: no
HEPES: 20 mM
L-glutamine: no
NaHCO3: yes
phenol red: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.

Application

RPMI-1640 Medium has been used

  • to obtain spleen and lymph nodes from mice
  • for the resuspension of Staphylococcus aureus strains in the phagocytosis assay
  • in the isolation of hepatitis B surface antigen-specific B-cell clones

Reconstitution

Supplement with 0.3 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blood polymorphonuclear leukocyte responses against Staphylococcus aureus in primiparous and pluriparous Lacaune and Santa Ines ewes
Marques M, et al.
Small Ruminant Research, 201, 106412-106412 (2021)
Kamila R Santos et al.
Vaccines, 9(8) (2021-08-29)
Staphylococcus aureus mastitis remains a major challenge for dairy farming. Here, 24 mice were immunized and divided into four groups: G1: control; G2: Granulocyte Macrophage Colony-Stimulating Factor (GM-CSF) DNA vaccine; G3: F0F1 ATP synthase subunit α (SAS), succinyl-diaminopimelate (SDD), and
Vitória M Silva et al.
BMC veterinary research, 17(1), 282-282 (2021-08-27)
The present study explored the viability of bovine milk macrophages, their intracellular production of reactive oxygen and nitrogen species (RONS), and their phagocytosis of Staphylococcus aureus, as well as the profile of lymphocytes, from healthy udder quarters and udder quarters
Ewerton de Souza Lima et al.
Frontiers in veterinary science, 8, 650021-650021 (2021-07-06)
The implications of bovine leukemia virus (BLV) on innate and adaptive immune responses have been widely investigated; however, the effects of BLV on mammary gland immunity require further investigation. The present study investigated the viability, phagocytic capacity, and intracellular production
Antonella Cerino et al.
PloS one, 10(4), e0125704-e0125704 (2015-04-30)
We describe the production and characterization of human monoclonal antibodies (mAb) specific for the major hepatitis B virus (HBV) S protein. The mAbs, two IgG1κ and one IgG1λ, were secreted by B-cell clones obtained from peripheral blood mononuclear cells (PBMC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service