Skip to Content
Merck
All Photos(1)

Key Documents

R3631

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule) antibody produced in sheep

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

sheep

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody subtype is the major serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids.

Application

Anti-Rabbit IgG (whole molecule) antibody produced in sheep has been used in competitive enzyme linked immunosorbent assay (ELISA).

Biochem/physiol Actions

IgG antibody defends bacterial, fungal and viral infections. Maternal IgG can cross placenta and offers immune defense for the neonate against infections.

Other Notes

Antibody adsorbed with human IgG.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a highly sensitive RIA for zoledronic acid, a new potent heterocyclic bisphosphonate, in human serum, plasma and urine
Legay F, et al.
Journal of Pharmaceutical and Biomedical Analysis, 30(4), 897-911 (2002)
Ganna Dmytrenko et al.
Journal of periodontal research, 59(2), 280-288 (2024-01-16)
The objective of the study was to evaluate the expression of oxytocin receptors in normal and inflamed gingiva, as well as the effects of systemic administration of oxytocin in bone loss and gum inflammatory mediators in a rat model of
C W Gray et al.
Methods in molecular medicine, 32, 217-228 (2000-01-01)
A detailed understanding of the biochemical events leading to the proteolytic excision of the β-amyloid peptide (A β) from the amyloid precursor protein (APP) has eluded many researchers. This is largely because the measurement of the various APP processing products
IL-21-induced isotype switching to IgG and IgA by human naive B cells is differentially regulated by IL-4
Avery DT, et al.
Journal of Immunology, 181(3), 1767-1779 (2008)
Placental transfer of IgG subclasses in a Japanese population.
Hashira S, et al.
Pediatrics International, 42(4) (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service