R352-05A
Rat Pulmonary Artery Smooth Muscle Cells: RPASMC, adult
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
rat pulmonary artery (healthy adult)
packaging
pkg of 500,000 cells
manufacturer/tradename
Cell Applications, Inc
growth mode
Adherent
morphology
smooth muscle
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cardiovascular diseases
shipped in
dry ice
storage temp.
−196°C
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Rat Pulmonary Artery Smooth Muscle Cells (RPASMC) provide a useful in vitro model to study cellular and molecular mechanisms of pulmonary hypertension characterized by hyperproliferation of pulmonary artery smooth muscle cells.Characterization: positive for smooth muscle cell specific alpha-actin expression.
Rat Pulmonary Artery Smooth Muscle Cells (RPASMC) provide a useful in vitro model to study cellular and molecular mechanisms of pulmonary hypertension characterized by hyperproliferation of pulmonary artery smooth muscle cells.Characterization: positive for smooth muscle cell specific alpha-actin expression.
Cell Line Origin
Artery
Application
Cell growth, signaling, receptor transactivation and suppression, vascular remodeling.
Components
Rat Smooth Muscle Cell Basal Medium that contains 10% FBS and 10% DMSO
Quality
Each lot was tested for proper morphology, Population Doublings, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.
Preparation Note
- 2nd passage, >500,000 cells in Rat Smooth Muscle Cell Basal Medium that contains 10% FBS and 10% DMSO
- Can be cultured at least 12 doublings
Subculture Routine
Please refer to the RPASMC Culture Protocol.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service