Skip to Content
Merck
All Photos(1)

Key Documents

R2533

Sigma-Aldrich

RS-127445 hydrochloride

≥98% (HPLC), powder

Synonym(s):

2-Amino-4-(4-fluoronaphth-1-yl)-6-isopropylpyrimidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16FN3 · HCl
CAS Number:
Molecular Weight:
317.79
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥20 mg/mL

originator

Roche

storage temp.

2-8°C

SMILES string

Cl.CC(C)c1cc(nc(N)n1)-c2ccc(F)c3ccccc23

InChI

1S/C17H16FN3.ClH/c1-10(2)15-9-16(21-17(19)20-15)13-7-8-14(18)12-6-4-3-5-11(12)13;/h3-10H,1-2H3,(H2,19,20,21);1H

InChI key

MKJPYBJBPRFMHL-UHFFFAOYSA-N

Biochem/physiol Actions

RS-127445 is a selective; high affinity; orally bioavailable serotonin 5-HT2B receptor antagonist. RS-127445 was found to have nanomolar affinity for the 5-HT2B receptor (pKi=9.5) and 1,000 fold selectivity for this receptor as compared to numerous other receptor and ion channel binding sites.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Chan Chang et al.
Cell communication and signaling : CCS, 22(1), 266-266 (2024-05-14)
Glioblastoma (GBM) is a type of brain cancer categorized as a high-grade glioma. GBM is characterized by limited treatment options, low patient survival rates, and abnormal serotonin metabolism. Previous studies have investigated the tumor suppressor function of aldolase C (ALDOC)
Maximilian Breuer et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 39(4), 498-512 (2024-03-13)
Mutations in the Chromodomain helicase DNA-binding protein 7 - coding gene (CHD7) cause CHARGE syndrome (CS). Although craniofacial and skeletal abnormalities are major features of CS patients, the role of CHD7 in bone and cartilage development remain largely unexplored. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service