Skip to Content
Merck
All Photos(3)

Key Documents

P8891

Sigma-Aldrich

Propantheline bromide

≥97% (TLC), powder

Synonym(s):

(2-Hydroxyethyl)diisopropylmethylammonium bromide xanthene-9-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C23H30NO3Br
CAS Number:
Molecular Weight:
448.39
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (TLC)

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL

originator

Shire

SMILES string

[Br-].CC(C)[N+](C)(CCOC(=O)C1c2ccccc2Oc3ccccc13)C(C)C

InChI

1S/C23H30NO3.BrH/c1-16(2)24(5,17(3)4)14-15-26-23(25)22-18-10-6-8-12-20(18)27-21-13-9-7-11-19(21)22;/h6-13,16-17,22H,14-15H2,1-5H3;1H/q+1;/p-1

InChI key

XLBIBBZXLMYSFF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits action of acetylcholine at post-ganglionic synapses; muscarinic acetylcholine receptor antagonist; antispasmodic; reduces gastric acid secretion.

Features and Benefits

This compound was developed by Shire. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Alaranta et al.
British journal of pharmacology, 101(2), 472-476 (1990-10-01)
1. The relative potency in blocking the nicotine-induced relaxation of the bovine retractor penis muscle (BRP) was estimated for 12 drugs known to have ganglion-blocking properties. 2. The order of potency of the drugs studied was mecamylamine greater than chlorisondamine
P Alberts
The Journal of pharmacology and experimental therapeutics, 274(1), 458-468 (1995-07-01)
The experiments were done to investigate the presence and subtype of functionally presynaptic muscarinic receptors in cholinergic nerves of the guinea pig urinary bladder. Bladder strips were incubated with 3H-choline and superfused with Tyrode's solution containing eserine. Secreted 3H-acetylcholine was
F Xavier Donadeu et al.
Biology of reproduction, 91(5), 111-111 (2014-09-26)
Several aspects of equine ovarian physiology are unique among domestic species. Moreover, follicular growth patterns are very similar between horses and humans. This study aimed to characterize, for the first time, global gene expression profiles associated with growth and preovulatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service