Skip to Content
Merck
All Photos(3)

Key Documents

P7561

Sigma-Aldrich

Phentolamine methanesulfonate salt

≥98% (TLC), powder

Synonym(s):

2-(N-[m-Hydroxyphenyl]-p-toluidinomethyl)imidazoline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H19N3O · CH3SO3H
CAS Number:
Molecular Weight:
377.46
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

180-182 °C (lit.)

solubility

ethanol: 26 mg/mL
H2O: 50 mg/mL

originator

Novartis

SMILES string

CS(O)(=O)=O.Cc1ccc(cc1)N(CC2=NCCN2)c3cccc(O)c3

InChI

1S/C17H19N3O.CH4O3S/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;1-5(2,3)4/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H3,(H,2,3,4)

InChI key

OGIYDFVHFQEFKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α-Adrenergic receptor antagonist; peripheral vasodilator.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jérémie Joffre et al.
Critical care medicine, 49(3), e315-e326 (2021-01-23)
Catecholaminergic vasopressors are the cornerstone of circulatory shock management. Nevertheless, catecholamines have problematic side effects, arousing a growing interest in noncatecholaminergic agents such as vasopressin or angiotensin-II. However, their respective effects on sepsis-associated microvascular endothelial dysfunction such as permeability or
Amanda M Wahl et al.
The Journal of physiology, 601(20), 4539-4556 (2023-09-19)
Currently, all salivary ducts (intercalated, striated and collecting) are assumed to function broadly in a similar manner, reclaiming ions that were secreted by the secretory acinar cells while preserving fluid volume and delivering saliva to the oral cavity. Nevertheless, there
Kazuhisa Honda et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 163(3-4), 260-264 (2012-08-23)
We investigated the mechanism underlying central glucagon-induced hyperglycemia and anorexia in chicks. Male 8-day-old chicks (Gallus gallus) were used in all experiments. Intracerebroventricular administration of glucagon in chicks induced hyperglycemia and anorexia from 30 min after administration. However, the plasma
Zachary Barrett-O'Keefe et al.
Clinical science (London, England : 1979), 124(6), 413-422 (2012-09-19)
Aging is characterized by increased sympatho-excitation, expressed through both the α-adrenergic and RAAS (renin-angiotensin-aldosterone) pathways. Although the independent contribution of these two pathways to elevated vasoconstriction with age may be substantial, significant cross-talk exists that could produce potentiating effects. To
Kangbin Zhou et al.
Canadian journal of physiology and pharmacology, 90(8), 1075-1085 (2012-07-13)
Metabolic syndrome (MS) is a combination of medical disorders that increase the risk of developing cardiovascular disease and diabetes. MS is associated with obesity, increased blood pressure, hyperlipidemia, and hyperglycemia. This study was designed to investigate the pharmacological profile of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service