Skip to Content
Merck
All Photos(3)

Key Documents

P3150

Sigma-Aldrich

Poly(Lys, Phe) 1:1 hydrobromide

mol wt 20,000-50,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

form

powder

Quality Level

feed ratio

Lys:Phe (1:1)

mol wt

20,000-50,000

color

white to off-white

application(s)

cell analysis

storage temp.

−20°C

Application

Poly (Lys, Phe) 1:1 hydrobromide has been used as a surfactant to bind negatively charged plasmid DNA. It has also been used to study its interaction with solvatochromic fluorescent dyes.

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of alternating poly(Lys-Phe) and comparison with the random copolymer poly(Lys 51, Phe 49).
G Seipke et al.
Biopolymers, 13(8), 1621-1633 (1974-01-01)
An Environmentally Sensitive Fluorescent Dye as a Multidimensional Probe of Amyloid Formation
Emma V. Yates
The Journal of Physical Chemistry, 120 (9), 2087?2094-2087?2094 (2016)
Helen Sjögren et al.
Biophysical chemistry, 116(1), 11-21 (2005-05-25)
The transition from alpha-helix to random coil of the titrating polyamino acid co-poly-L-(lysine, phenylalanine), (p-(Lys,Phe)), has been investigated as a function of pH and ionic strength in aqueous solution and at the air-water interface by means of circular dichroism (CD)
Tae Woo Kim et al.
International journal of pharmaceutics, 295(1-2), 35-45 (2005-04-26)
To enhance the in vitro and in vivo transfection activity of the cationic lipid emulsion (LE), three natural polycations, protamine sulfate (PS), poly-L-lysine and spermine, were selected as DNA condensing active agents. Formation of the LE/polycation/DNA ternary complexes was identified
Optimising DNA binding to carbon nanotubes by non-covalent methods
Vanesa Sanz
Carbon, 49(5), 1775-1781 (2011)

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service