Skip to Content
Merck
All Photos(1)

Key Documents

N9158

Sigma-Aldrich

N-(NBD-Aminolauroyl)ceramide

≥98%

Synonym(s):

C12-NBD Ceramide, N-[12-[(7-Nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-ceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H61N5O6
Molecular Weight:
659.90
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

solid

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(=O)CCCCCCCCCCCNc1ccc([N+]([O-])=O)c2nonc12

InChI

1S/C36H61N5O6/c1-2-3-4-5-6-7-8-9-10-12-15-18-21-24-33(43)31(29-42)38-34(44)25-22-19-16-13-11-14-17-20-23-28-37-30-26-27-32(41(45)46)36-35(30)39-47-40-36/h21,24,26-27,31,33,37,42-43H,2-20,22-23,25,28-29H2,1H3,(H,38,44)/b24-21+/t31-,33+/m0/s1

InChI key

SNOJCYCOPNIGIK-ULETYAGTSA-N

General description

Fluorescently labeled fatty acid sphingolipid membrane probe. NBD group excitation 460 nm, emission 534 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaalee Dworski et al.
Biochimica et biophysica acta, 1863(2), 386-394 (2016-12-05)
Acid Ceramidase Deficiency (Farber disease, FD) is an ultra-rare Lysosomal Storage Disorder that is poorly understood and often misdiagnosed as Juvenile Idiopathic Arthritis (JIA). Hallmarks of FD are accumulation of ceramides, widespread macrophage infiltration, splenomegaly, and lymphocytosis. The cytokines involved
Paola Palumbo et al.
International journal of molecular sciences, 20(12) (2019-06-23)
The relevance of nitric oxide synthase 2 (NOS2) as a prognostic factor in Glioblastoma Multiforme (GBM) malignancy is emerging. We analyzed the effect of NOS2 inhibitor 1400W on the autophagic flux and extracellular vesicle (EV) secretion in U87MG glioma cells.
Wataru Sakamoto et al.
Journal of lipid research, 59(11), 2116-2125 (2018-08-30)
Ceramidases hydrolyze ceramides into sphingosine and fatty acids, with sphingosine being further metabolized into sphingosine-1-phosphate (S1P); thus, ceramidases control the levels of these bioactive sphingolipids in cells and tissues. Neutral ceramidase (nCDase) is highly expressed in colorectal tissues, and a
Cassandra Suhrland et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(3), 1141-1156 (2019-08-25)
The bioactive sphingolipid ceramide has many important roles in cell signaling processes, particularly in signaling programmed cell death in cancer. However, ceramide levels are often impaired in multi-drug resistant and radiation resistant cancers due to the dysregulation of ceramide metabolism.
Anis Ahmad et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 31(2), 771-780 (2016-11-12)
The molecular mechanisms responsible for the development of proteinuria and glomerulosclerosis in radiation nephropathy remain largely unknown. Podocytes are increasingly recognized as key players in the pathogenesis of proteinuria in primary and secondary glomerular disorders. The lipid-modulating enzyme sphingomyelin phosphodiesterase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service