Skip to Content
Merck
All Photos(4)

Key Documents

N3139

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate

powder, BioReagent, suitable for cell culture

Synonym(s):

Triphosphopyridine nucleotide sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7NaO17P3 · xH2O
CAS Number:
Molecular Weight:
765.39 (anhydrous basis)
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

Assay

≥98% (HPLC)
≥98% (spectrophotometric assay)

form

powder

technique(s)

cell culture | mammalian: suitable

mp

175-178 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O)[C@H]([N+]2=CC(C(N)=O)=CC=C2)O[C@@H]1COP(OP(OC[C@H]3O[C@@H](N(C=N4)C5=C4C(N)=NC=N5)[C@H](OP(O)(O)=O)[C@@H]3O)([O-])=O)([O-])=O.O.[Na+]

InChI

1S/C21H28N7O17P3.Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39);;1H2/q;+1;/p-1/t10-,11-,13-,14-,15-,16-,20-,21-;;/m1../s1

InChI key

CYQLUFJYVTUUFN-WUEGHLCSSA-M

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate has been used in the purification and crystallization of CHORD–Sgt1 cytochrome b 5 reductases (CS–b5R). It has also been used as a supplement in Dulbecco′s modified Eagle′s medium (DMEM)/F-12 medium for direct hepatic reprogramming using mouse embryonic fibroblasts (MEFs).

Biochem/physiol Actions

Electron acceptor
Nicotinamide adenine dinucleotide phosphate (NADP) is an indispensable cofactor that participates in cellular redox reactions. It acts as a hydride-accepting coenzyme and participates in substrate oxidation reactions during metabolism.

Packaging

Packaged by solid weight.

Analysis Note

Meets or exceeds NRC specifications and is comparable to any commercial preparation available.

Other Notes

This is the common form of NADP. Do not confuse with 3′−NADP or α-NADP.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David R Benson et al.
Acta crystallographica. Section D, Structural biology, 75(Pt 7), 628-638 (2019-07-10)
Ncb5or (NADH-cytochrome b5 oxidoreductase), a cytosolic ferric reductase implicated in diabetes and neurological diseases, comprises three distinct domains, cytochrome b5 (b5) and cytochrome b5 reductase (b5R) domains separated by a CHORD-Sgt1 (CS) domain, and a novel 50-residue N-terminal region. Understanding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service