Skip to Content
Merck
All Photos(1)

Key Documents

M9651

Sigma-Aldrich

Maprotiline hydrochloride

>99% (HPLC), powder

Synonym(s):

9-(γ-Methylaminopropyl)-9,10-dihydro-9,10-ethanoanthracene hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N · HCl
CAS Number:
Molecular Weight:
313.86
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>99% (HPLC)

form

powder

color

white

solubility

H2O: 50 mg/mL

originator

Novartis

SMILES string

Cl.CNCCCC12CCC(c3ccccc13)c4ccccc24

InChI

1S/C20H23N.ClH/c1-21-14-6-12-20-13-11-15(16-7-2-4-9-18(16)20)17-8-3-5-10-19(17)20;/h2-5,7-10,15,21H,6,11-14H2,1H3;1H

InChI key

NZDMFGKECODQRY-UHFFFAOYSA-N

Gene Information

human ... SLC6A2(6530)

Looking for similar products? Visit Product Comparison Guide

Application

Maprotiline hydrochloride has been used as a inhibitor of norepinephrine uptake to study its effects on arresting L3055 cell proliferation.

Biochem/physiol Actions

Maprotiline is a tetracyclic agent. It possesses inhibitory effects against the uptake of norepinephrine by nerve cells. Maprotiline exhibits anti-depressant and sedative activity.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Hösli et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 13(8), 897-908 (1995-12-01)
Autoradiographic studies were made on the uptake of 3H-noradrenaline and 3H-serotonin in explant cultures and primary astrocyte cultures from various regions of rat central nervous system (cortex, cerebellum, locus coeruleus, nucleus raphé, spinal cord). In explant cultures from locus coeruleus
C C Beaufour et al.
Pharmacology, biochemistry, and behavior, 62(4), 591-599 (1999-04-20)
The effects of chronic antidepressants were investigated in an animal procedure for the study of anxiety and anxiolytics, the conditioned suppression of operant behavior in rats. In daily 18-min sessions, three periods of nonpunished lever pressing for food alternated with
The serotonin transporter (SLC6A4) is present in B-cell clones of diverse malignant origin: probing a potential antitumor target for psychotropics
Meredith E J, et al.
Faseb Journal, 19(9), 1187-1189 (2005)
Chemistry, pharmacology, pharmacokinetics, adverse effects, and efficacy of the antidepressant maprotiline hydrochloride
Wells B G and Gelenberg A J
Pharmacotherapy, 1(2), 121-138 (1981)
Murat Pinar et al.
Progress in neuro-psychopharmacology & biological psychiatry, 32(1), 135-139 (2007-09-11)
Agents such as clozapine, olanzapine and mirtazapine frequently trigger an increase in body weight. Though the mechanisms have not been thoroughly clarified, recent studies indicate a role for ghrelin in regulation of appetite and weight gain. We investigated the relation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service