Skip to Content
Merck
All Photos(1)

Documents

M1170

Sigma-Aldrich

Met-Arg-Phe-Ala acetate salt

≥90% (HPLC), suitable for MALDI-MS

Synonym(s):

L-methionyl-L-arginyl-L-phenylalanyl-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H37N7O5S
CAS Number:
Molecular Weight:
523.65
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Met-Arg-Phe-Ala acetate salt, ≥90% (HPLC)

Quality Level

Assay

≥90% (HPLC)

form

powder

technique(s)

MALDI-MS: suitable

color

white

storage temp.

−20°C

SMILES string

CSCCC(N)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc1ccccc1)C(=O)NC(C)C(O)=O

InChI

1S/C23H37N7O5S/c1-14(22(34)35)28-21(33)18(13-15-7-4-3-5-8-15)30-20(32)17(9-6-11-27-23(25)26)29-19(31)16(24)10-12-36-2/h3-5,7-8,14,16-18H,6,9-13,24H2,1-2H3,(H,28,33)(H,29,31)(H,30,32)(H,34,35)(H4,25,26,27)

InChI key

UWTASUKCZNGRDU-UHFFFAOYSA-N

Amino Acid Sequence

Met-Arg-Phe-Ala

Application

A marker in an unambiguous method for sequencing tetrapeptides using FAB, MI, and collisional activation spectra in combination.
Met-Arg-Phe-Ala acetate salt has been used as an internal standard in the mass determination of the compounds by electrospray ionization mass spectrometry (ESI-MS). It has also been used in comparing the laser energy used in nanostructure-initiator mass spectrometry (NIMS) and matrix-assisted laser desorption/ionization (MALDI).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Ponzetto et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1052, 34-42 (2017-03-28)
Recently, steroid hormones quantification in blood showed a promising ability to detect testosterone doping and interesting complementarities with the urinary module of the Athlete Biological Passport (ABP). In this work, an ultra-high pressure liquid chromatography-high-resolution mass spectrometry (UHPLC-HRMS) method was
Kulik, W. and Heerma, W.
Biomedical Mass Spectrometry, 20(9), 553-553 (1991)
Kin-Sing Wong et al.
Journal of proteome research, 19(3), 1196-1208 (2020-02-14)
Recent advances in label-free quantitative proteomics may support its application in identifying and monitoring biomarkers for the purpose of doping control in equine sports. In this study, we developed a workflow of label-free quantitative proteomics to propose plasma protein biomarkers
Marina López-García et al.
Journal of chromatography. A, 1540, 21-30 (2018-02-06)
A new analytical method based on ultra-high performance liquid chromatography (UHPLC) coupled to Orbitrap high resolution mass spectrometry (Orbitrap-HRMS) has been developed for the determination of steroid hormones (hydrocortisone, cortisone, progesterone, prednisone, prednisolone, testosterone, melengesterol acetate, hydrocortisone-21-acetate, cortisone-21-acetate, testosterone propionate
Kin-Sing Wong et al.
Journal of chromatography. A, 1478, 35-42 (2016-12-05)
Growth hormone (GH), or somatotropin, is a protein that may enhance physical performance and facilitate growth and wound healing. For this reason, growth hormones and their recombinant analogues are prohibited in human sports by the World Anti-Doping Agency (WADA) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service