Skip to Content
Merck
All Photos(2)

Key Documents

L1754

Sigma-Aldrich

Lipase from Candida rugosa

Type VII, ≥700 unit/mg solid

Synonym(s):

Triacylglycerol acylhydrolase, Triacylglycerol lipase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fungus (candida rugosa)

type

Type VII

form

lyophilized

specific activity

≥700 unit/mg solid

storage condition

(Tightly closed. Dry)

technique(s)

cell based assay: suitable

color

beige
white

solubility

water: slightly soluble

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Research area: Cell Structure

Lipases are found in pancreatic secretions. This class of enzymes contains α and β hydrolase folds. Candida rugosa produces multiple lipase isoenzymes that have 80% sequence homology.
This product contains lactose as a carrier.

Application

Lipase from Candida rugosa has been used:
  • to synthesize dextran fatty acid esters
  • to study the effect of lipase on conjugated linoleic acid (CLA1 and CLA2) production in the presence of sunflower oil and castor oil
  • to prepare pH-imprinted enzyme for lipase-catalyzed transesterification of dextran T-40 with vinyl decanoate

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

Lipases are enzymes and biocatalysts that catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids. They are useful for biotransformation reactions. It possesses broad substrate tolerance and is highly stereo- and regioselective. Lipases mediate lipid transport. Elevated levels of serum lipase are observed in pancreatitis. Lipases are the preferred catalysts for long-chain fatty acids. Lipases can be used in a wide range of solvents, in both aqueous and non-aqueous media, and thus have found use in applications like organic synthesis.
Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).
Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Unit Definition

One unit will hydrolyze 1.0 microequivalent of fatty acid from a triglyceride in 1 hr at pH 7.2 at 37 °C. (This is equivalent to approx. 10 microliters of CO2 in 30 minutes.)

enzyme

Product No.
Description
Pricing

substrate

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashok Kumar et al.
Biological procedures online, 18, 2-2 (2016-01-15)
Lipases are industrial biocatalysts, which are involved in several novel reactions, occurring in aqueous medium as well as non-aqueous medium. Furthermore, they are well-known for their remarkable ability to carry out a wide variety of chemo-, regio- and enantio-selective transformations.
Taresh P Khobragade et al.
Frontiers in bioengineering and biotechnology, 9, 757062-757062 (2021-10-26)
Herein, we report the development of a multi-enzyme cascade using transaminase (TA), esterase, aldehyde reductase (AHR), and formate dehydrogenase (FDH), using benzylamine as an amino donor to synthesize the industrially important compound sitagliptin intermediate. A panel of 16 TAs was
Marco Filice et al.
Nature protocols, 7(10), 1783-1796 (2012-09-08)
This protocol describes the regioselective deprotection of single hydroxyls in peracetylated monosaccharides and disaccharides by enzymatic or chemoenzymatic strategies. The introduction of a one-pot enzymatic step by using immobilized biocatalysts obviates the requirement to carry out tedious workups and time-consuming
Enzymatic synthesis of oligo- and polysaccharide fatty acid esters.
van den Broek, et al.
Carbohydrate Polymers, 93, 65-72 (2013)
Lipase
Pirahanchi Y and SharmaS
Biochemistry (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service