Skip to Content
Merck
All Photos(1)

Key Documents

L1645

Sigma-Aldrich

Anti-Human Lambda Light Chain (Bound and Free) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:2,500

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Immunoglobulins (Ig) are produced by the B cells and play crucial role in host humoral immune response. A typical immunoglobulin consists of two heavy chains and two light chains. The two light chains are either κ or the λ chain. The B cells express one type of light chain during their life time.
Lambda (λ) light chain comprises an amino-terminal variable region and carboxy-terminal constant region. λ chain is biologically critical and constitutes up to 30 to 40% of human immunoglobulin (Ig) molecules. The human λ locus is mapped to the chromosome location 22.

Application

Anti-Human Lambda Light Chain (Bound and Free) antibody has been used in surface plasmon resonance assay.[1]
Anti-Human Lambda Light Chain (bound and free) antibody produced in goat has been used in indirect enzyme linked immuno sorbent assay (ELISA).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.For extended storage, solution may be frozen inworking aliquots. Repeated freezing and thawing is notrecommended. Storage in "frost-free" freezers is notrecommended.If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and functional properties of human lambda-light-chain variable-region subgroups.
A Solomon et al.
Clinical and diagnostic laboratory immunology, 2(4), 387-394 (1995-07-01)
B-cell development
Hematology (2018)
Adam W Clarke et al.
mAbs, 10(4), 664-677 (2018-02-14)
TL1A is an attractive therapeutic target for the treatment of mucosal inflammation associated with inflammatory bowel disease (IBD) and asthma. Blockade of the TL1A pathway has been shown to reduce inflammatory responses while leaving baseline immunity intact, and to be
Jinhao Bi et al.
Emerging microbes & infections, 12(1), 2149351-2149351 (2022-12-02)
Marburg virus disease (MVD) is a lethal viral haemorrhagic fever caused by Marburg virus (MARV) with a case fatality rate as high as 88%. There is currently no vaccine or antiviral therapy approved for MVD. Due to high variation among
Jerry A Katzmann et al.
Clinical chemistry, 48(9), 1437-1444 (2002-08-27)
The detection of monoclonal free light chains (FLCs) is an important diagnostic aid for a variety of monoclonal gammopathies and is especially important in light-chain diseases, such as light-chain myeloma, primary systemic amyloidosis, and light-chain-deposition disease. These diseases are more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service