Skip to Content
Merck
All Photos(1)

Key Documents

L0521

Sigma-Aldrich

Lipoxin A4

ethanol solution

Synonym(s):

(5S,6R,15S)-Trihydroxy-(7E,9E,11Z,13E)-Eicosatetraenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

form

ethanol solution

Quality Level

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)\C=C\C=C/C=C/C=C/[C@@H](O)[C@@H](O)CCCC(O)=O

InChI

1S/C20H32O5/c1-2-3-8-12-17(21)13-9-6-4-5-7-10-14-18(22)19(23)15-11-16-20(24)25/h4-7,9-10,13-14,17-19,21-23H,2-3,8,11-12,15-16H2,1H3,(H,24,25)/b6-4-,7-5+,13-9+,14-10+/t17-,18+,19-/m0/s1

InChI key

IXAQOQZEOGMIQS-SSQFXEBMSA-N

Biochem/physiol Actions

Lipoxin A4 (LXA4) is synthesized from arachidonic acid and is an endogenous lipoxygenase-derived eicosanoid mediator. It is a potent human protein kinase C activator. It inhibits cytotoxicity of natural killer cells. LXA4 regulates the immune system and inflammation. In the brain, it modulates neuronal signaling and slow wave sleep. LXA4 binds to cannabinoid receptors. LXA4 plays a key role in the maintenance of endometrium and reproductive function. Depletion of LXA4 contributes to the pathophysiology of cystic fibrosis (CF).

Packaging

Packaged under argon.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K C Nicolaou et al.
Biochimica et biophysica acta, 1003(1), 44-53 (1989-05-15)
Addition of (15S)-hydroxy-5,8,11-cis-13-trans-eicosatetraenoic acid (15-HETE) and the ionophore A23187 (2.5 microM) to human neutrophils led to the formation of both lipoxin A4 and lipoxin B4 as well as a novel 5,6,15-trihydroxyeicosatetraenoic acid. The new compound was identified using an improved
Physiological impact of abnormal lipoxin A4 production on cystic fibrosis airway epithelium and therapeutic potential.
Higgins G, et al.
BioMed Research International, 2015(3), 439-439 (2015)
Anti-inflammatory lipoxin A4 is an endogenous allosteric enhancer of CB1 cannabinoid receptor.
Pamplona FA, et al.
Proceedings of the National Academy of Sciences of the USA, 109(51), 21134-21139 (2012)
The role of lipoxin A 4 in endometrial biology and endometriosis.
Canny GO and Lessey BA
Mucosal Immunology, 6(3), 439-439 (2013)
L Habouri et al.
Osteoarthritis and cartilage, 25(10), 1719-1728 (2017-07-12)
12/15-Lipoxygenase (12/15-LOX) catalyzes the generation of various anti-inflammatory lipid mediators, and has been implicated in several inflammatory and degenerative diseases. However, there is currently no evidence that 12/15-LOX has a role in osteoarthritis (OA). The aim of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service