Skip to Content
Merck
All Photos(1)

Key Documents

I8273

Sigma-Aldrich

Anti-Interleukin-2 antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-IL-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

neutralization: suitable
western blot: 1 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL2(3558)

Looking for similar products? Visit Product Comparison Guide

General description

Interleukin-2 (IL-2) is a cytokine produced by activated T cells in response to antigen stimulation. IL-2 is important for proliferation of lymphocytes, natural killer cells and macrophages and in the differentiation of CD4+ cells into Th1 and Th2 effector cells. The effects of IL-2 are mediated by the signalling activated by its binding to IL-2 receptor (IL-2 R). IL-2 activates key signalling pathways such as MAPK, JAK/STAT as well as PI3K/Akt. Activation of these pathways modulates production of inflammatory cytokines, survival and progression through cell cycle
Monoclonal Anti-Interleukin-2 recognizes human Interleukin-2. It also neutralizes recombinant, mouse IL-2, but requires 30-50 times the antibody concentration required to neutralize human IL-2.

Specificity

The antibody neutralizes the biological activity of recombinant human IL-2.

Immunogen

recombinant human interleukin-2 expressed in Escherichia coli.

Application

Anti-interleukin-2 antibody is suitable for IL-2 neutralization assays.

Physical form

Lyophilized from a 0.2 μm filtered solution of phosphate buffered saline containing carbohydrates.

Preparation Note

Purified using protein G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew A Williams et al.
Nature, 441(7095), 890-893 (2006-06-17)
Although interleukin-2 (IL-2) was initially characterized as the primary T-cell growth factor following in vitro activation, less is known about its role in shaping T-cell responses to acute infections in vivo. The use of IL-2- or IL-2-receptor-deficient mice is problematic
B H Nelson et al.
Advances in immunology, 70, 1-81 (1998-10-02)
Studies of the biology of the IL-2 receptor have played a major part in establishing several of the fundamental principles that govern our current understanding of immunology. Chief among these is the contribution made by lymphokines to regulation of the
Katrina K Hoyer et al.
Immunological reviews, 226, 19-28 (2009-01-24)
Interleukin-2 (IL-2) has multiple, sometimes opposing, functions during an inflammatory response. It is a potent inducer of T-cell proliferation and T-helper 1 (Th1) and Th2 effector T-cell differentiation and provides T cells with a long-lasting competitive advantage resulting in the
Hans Dooms et al.
Journal of immunology (Baltimore, Md. : 1950), 172(10), 5973-5979 (2004-05-07)
The acquisition of long-term survival potential by activated T lymphocytes is essential to ensure the successful development of a memory population in the competitive environment of the lymphoid system. The factors that grant competitiveness for survival to primed T cells
Arian Laurence et al.
Immunity, 26(3), 371-381 (2007-03-17)
Recent work has identified a new subset of effector T cells that produces interleukin (IL)-17 known as T helper 17 (Th17) cells, which is involved in the pathophysiology of inflammatory diseases and is thought to be developmentally related to regulatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service