Skip to Content
Merck
All Photos(4)

Key Documents

HPA020433

Sigma-Aldrich

Anti-PLS3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-T-plastin, Anti-plastin 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PAKFSLVGIGGQDLNDGNQTLTLALVWQLMRRYTLNVLEDLGDGQKANDDIIVNWVNRTLSEAGKSTSIQSFKDKTISSSLAVVDLIDAIQPGCINYDLVKSGNLTEDDKHNNAKYAVSMARRIGARVYALPEDLVEVKPGAPHR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLS3(5358)

General description

Plastin 3 (PLS3) is expressed in epithelial cells. The gene encoding it is localized on human chromosome Xq23.

Immunogen

plastin 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Plastin 3 (PLS3) imparts the bundled nature to actin filaments and cross-links to them, thereby helping in cell motility. In circulating tumor cells, it stimulates epithelial-mesenchymal transition (EMT) through the transforming growth factor-β (TGF-β) pathway. Mutation in the PLS3 gene leads to X-linked low-turnover osteoporosis and the protein has been found to be expressed in breast cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82421

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine M Laine et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 30(3), 510-518 (2014-09-12)
Genetic factors play an important role in the development of osteoporosis. Several monogenic forms of osteoporosis have been recognized, most recently an X-chromosomal form resulting from mutations in the gene encoding plastin 3 (PLS3). PLS3 is a protein involved in
H Ueo et al.
British journal of cancer, 112(9), 1519-1526 (2015-04-17)
Identification of promising biomarkers that predict the prognosis of patients with breast cancer is needed. In this study, we hypothesised that the expression of the epithelial-mesenchymal transition-related biomarker plastin3 (PLS3) in peripheral blood could be a prognostic factor in breast
Keishi Sugimachi et al.
Annals of surgical oncology, 21(11), 3680-3690 (2013-11-13)
Plastin-3 (PLS3) is a novel marker for circulating tumor cells (CTCs) in colorectal cancer (CRC). We sought to investigate the mechanisms mediating the aberrant expression of PLS3, the role of PLS3 in the epithelial-mesenchymal transition (EMT), and its association with
Christine L Jones et al.
The Journal of investigative dermatology, 132(8), 2042-2049 (2012-04-13)
T-plastin (PLS3) is an actin-bundling protein normally expressed in epithelial cells but absent in cells of hematopoietic origin. Aberrant PLS3 expression has been demonstrated in lymphocytes from Sézary syndrome (SS) patients and has been proposed as a biomarker for SS;
Cécilia Brun et al.
PloS one, 9(9), e104700-e104700 (2014-09-17)
Cutaneous wound healing requires keratinocyte proliferation, migration and differentiation to restore the barrier function of the skin. The calcineurin/nuclear factor of activated-T-cell (NFAT) signaling pathway has been recently shown to be involved in keratinocyte growth, differentiation and migration. It is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service